Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 32973168-32973429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAAP24
Entrez Gene ID
91442
Gene Name
Fanconi anemia core complex associated protein 24
Gene Aliases
C19orf40
UniGene
Hs.579899
Gene Chromosome Location
Chr.19: 32972212-32977495 on Build GRCh38
Transcripts
NM_001300978.1 NM_152266.4 XM_005259393.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4469450 c.15C>T p.P5P Chr.19: 32973211-32973211 on Build GRCh38
1641119 c.27G>A p.T9T Chr.19: 32973223-32973223 on Build GRCh38
4076736 c.67G>T p.E23* Chr.19: 32973263-32973263 on Build GRCh38
5742673 c.79G>A p.G27R Chr.19: 32973275-32973275 on Build GRCh38
4076737 c.69G>A p.E23E Chr.19: 32973265-32973265 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759255098 Chr.19: 32973232-32973232 on Build GRCh38
rs913701148 Chr.19: 32973355-32973355 on Build GRCh38
rs777882240 Chr.19: 32973199-32973199 on Build GRCh38
rs766438613 Chr.19: 32973324-32973324 on Build GRCh38
rs767627178 Chr.19: 32973253-32973253 on Build GRCh38
rs949386353 Chr.19: 32973306-32973306 on Build GRCh38
rs73037453 Chr.19: 32973319-32973319 on Build GRCh38
rs200046285 Chr.19: 32973288-32973288 on Build GRCh38
rs769628150 Chr.19: 32973307-32973307 on Build GRCh38
rs368480238 Chr.19: 32973246-32973246 on Build GRCh38
rs775238035 Chr.19: 32973312-32973312 on Build GRCh38
rs756420548 Chr.19: 32973260-32973260 on Build GRCh38
rs749606882 Chr.19: 32973197-32973197 on Build GRCh38
rs747509992 Chr.19: 32973386-32973386 on Build GRCh38
rs776547102 Chr.19: 32973230-32973230 on Build GRCh38
rs979561969 Chr.19: 32973231-32973231 on Build GRCh38
rs760413240 Chr.19: 32973223-32973223 on Build GRCh38
rs754027420 Chr.19: 32973332-32973332 on Build GRCh38
rs780149743 Chr.19: 32973261-32973261 on Build GRCh38
rs576051776 Chr.19: 32973274-32973274 on Build GRCh38
rs530097367 Chr.19: 32973217-32973217 on Build GRCh38
rs774998677 Chr.19: 32973202-32973202 on Build GRCh38
rs200833742 Chr.19: 32973273-32973273 on Build GRCh38
rs759032259 Chr.19: 32973384-32973384 on Build GRCh38
rs371874659 Chr.19: 32973259-32973259 on Build GRCh38
rs529898862 Chr.19: 32973337-32973337 on Build GRCh38
rs771726752 Chr.19: 32973209-32973209 on Build GRCh38
rs931407939 Chr.19: 32973352-32973352 on Build GRCh38
rs751459842 Chr.19: 32973321-32973321 on Build GRCh38
rs900861352 Chr.19: 32973222-32973222 on Build GRCh38
rs754020925 Chr.19: 32973272-32973272 on Build GRCh38
rs764976500 Chr.19: 32973244-32973244 on Build GRCh38
rs149750443 Chr.19: 32973303-32973303 on Build GRCh38
rs748409624 Chr.19: 32973348-32973348 on Build GRCh38
rs780510110 Chr.19: 32973338-32973339 on Build GRCh38
rs987176140 Chr.19: 32973385-32973385 on Build GRCh38
rs367939215 Chr.19: 32973289-32973289 on Build GRCh38
rs773336436 Chr.19: 32973348-32973349 on Build GRCh38
rs753106057 Chr.19: 32973348-32973348 on Build GRCh38
rs536928664 Chr.19: 32973347-32973348 on Build GRCh38
rs770370535 Chr.19: 32973391-32973391 on Build GRCh38
rs748623265 Chr.19: 32973203-32973203 on Build GRCh38
rs770484990 Chr.19: 32973229-32973229 on Build GRCh38
rs34151845 Chr.19: 32973225-32973225 on Build GRCh38
rs755103549 Chr.19: 32973226-32973226 on Build GRCh38
rs760671671 Chr.19: 32973322-32973322 on Build GRCh38
rs1051873950 Chr.19: 32973262-32973262 on Build GRCh38
rs762947159 Chr.19: 32973313-32973313 on Build GRCh38
rs201531119 Chr.19: 32973275-32973275 on Build GRCh38
rs770407899 Chr.19: 32973296-32973296 on Build GRCh38
rs888771522 Chr.19: 32973269-32973269 on Build GRCh38
rs776179788 Chr.19: 32973297-32973297 on Build GRCh38
rs1032542805 Chr.19: 32973233-32973233 on Build GRCh38
rs148264788 Chr.19: 32973342-32973342 on Build GRCh38
rs772673556 Chr.19: 32973282-32973282 on Build GRCh38
rs762858523 Chr.19: 32973252-32973252 on Build GRCh38
rs759267817 Chr.19: 32973340-32973340 on Build GRCh38

View Full Product Details