Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6677955-6678212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3
Entrez Gene ID
718
Gene Name
complement component 3
Gene Aliases
AHUS5, ARMD9, ASP, C3a, C3b, CPAMD1, HEL-S-62p
UniGene
Hs.529053
Gene Chromosome Location
Chr.19: 6677835-6720682 on Build GRCh38
Transcripts
NM_000064.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6310012 c.4913A>C p.Q1638P Chr.19: 6677961-6677961 on Build GRCh38
5857027 c.4893G>T p.W1631C Chr.19: 6677981-6677981 on Build GRCh38
7257976 c.4846C>T p.P1616S Chr.19: 6678156-6678156 on Build GRCh38
6615967 c.4821C>T p.L1607L Chr.19: 6678181-6678181 on Build GRCh38
4581552 c.4843A>G p.K1615E Chr.19: 6678159-6678159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373087196 Chr.19: 6677974-6677974 on Build GRCh38
rs770052443 Chr.19: 6678072-6678072 on Build GRCh38
rs17030 Chr.19: 6677978-6677978 on Build GRCh38
rs1803223 Chr.19: 6677996-6677996 on Build GRCh38
rs570044337 Chr.19: 6678071-6678071 on Build GRCh38
rs958247533 Chr.19: 6678074-6678075 on Build GRCh38
rs1012252412 Chr.19: 6678075-6678075 on Build GRCh38
rs1023638329 Chr.19: 6678084-6678084 on Build GRCh38
rs931697966 Chr.19: 6677986-6677986 on Build GRCh38
rs2230210 Chr.19: 6678019-6678019 on Build GRCh38
rs879127151 Chr.19: 6678151-6678151 on Build GRCh38
rs879166258 Chr.19: 6678150-6678151 on Build GRCh38
rs3189774 Chr.19: 6678169-6678169 on Build GRCh38
rs766517800 Chr.19: 6677999-6677999 on Build GRCh38
rs536092913 Chr.19: 6678093-6678093 on Build GRCh38
rs888299834 Chr.19: 6677980-6677980 on Build GRCh38
rs539822147 Chr.19: 6678005-6678005 on Build GRCh38
rs911645126 Chr.19: 6678181-6678181 on Build GRCh38
rs770287028 Chr.19: 6678074-6678074 on Build GRCh38
rs772905898 Chr.19: 6678124-6678133 on Build GRCh38
rs747280056 Chr.19: 6678044-6678044 on Build GRCh38
rs185428134 Chr.19: 6678149-6678149 on Build GRCh38
rs910777622 Chr.19: 6678063-6678075 on Build GRCh38
rs200816772 Chr.19: 6677987-6677987 on Build GRCh38
rs748416799 Chr.19: 6678140-6678140 on Build GRCh38
rs746415216 Chr.19: 6678057-6678086 on Build GRCh38
rs781676250 Chr.19: 6678137-6678137 on Build GRCh38
rs775140871 Chr.19: 6677993-6677993 on Build GRCh38
rs776406027 Chr.19: 6678182-6678182 on Build GRCh38
rs344556 Chr.19: 6678104-6678104 on Build GRCh38
rs150537373 Chr.19: 6678175-6678175 on Build GRCh38
rs374509630 Chr.19: 6678103-6678103 on Build GRCh38
rs113627602 Chr.19: 6678102-6678102 on Build GRCh38
rs753349401 Chr.19: 6678017-6678017 on Build GRCh38
rs374395215 Chr.19: 6678121-6678121 on Build GRCh38
rs879114238 Chr.19: 6678146-6678146 on Build GRCh38
rs751987860 Chr.19: 6678129-6678129 on Build GRCh38
rs370787795 Chr.19: 6678041-6678041 on Build GRCh38
rs771728904 Chr.19: 6678124-6678131 on Build GRCh38
rs764795400 Chr.19: 6678117-6678117 on Build GRCh38
rs139457470 Chr.19: 6678178-6678178 on Build GRCh38
rs773766353 Chr.19: 6678108-6678108 on Build GRCh38
rs376167337 Chr.19: 6678066-6678066 on Build GRCh38
rs1803220 Chr.19: 6678166-6678166 on Build GRCh38
rs765001904 Chr.19: 6678135-6678135 on Build GRCh38
rs546786971 Chr.19: 6678100-6678100 on Build GRCh38
rs761423874 Chr.19: 6678113-6678113 on Build GRCh38
rs988674147 Chr.19: 6678090-6678090 on Build GRCh38
rs749266657 Chr.19: 6678162-6678162 on Build GRCh38
rs112179814 Chr.19: 6678024-6678024 on Build GRCh38
rs1032790084 Chr.19: 6678073-6678073 on Build GRCh38
rs141661874 Chr.19: 6678164-6678164 on Build GRCh38
rs747630475 Chr.19: 6678119-6678126 on Build GRCh38
rs780527356 Chr.19: 6678042-6678042 on Build GRCh38
rs773390079 Chr.19: 6678101-6678101 on Build GRCh38
rs775112459 Chr.19: 6678152-6678152 on Build GRCh38
rs755235514 Chr.19: 6678050-6678050 on Build GRCh38
rs893904061 Chr.19: 6678074-6678074 on Build GRCh38
rs766732348 Chr.19: 6678128-6678128 on Build GRCh38
rs879224714 Chr.19: 6678028-6678028 on Build GRCh38
rs763838624 Chr.19: 6678107-6678107 on Build GRCh38
rs200817637 Chr.19: 6678180-6678180 on Build GRCh38
rs774972918 Chr.19: 6677995-6677995 on Build GRCh38
rs759278730 Chr.19: 6678096-6678096 on Build GRCh38
rs371253947 Chr.19: 6678109-6678109 on Build GRCh38
rs755252899 Chr.19: 6678134-6678134 on Build GRCh38
rs971175399 Chr.19: 6678088-6678088 on Build GRCh38
rs1042065889 Chr.19: 6678045-6678045 on Build GRCh38
rs980807662 Chr.19: 6678064-6678064 on Build GRCh38
rs374196546 Chr.19: 6678060-6678060 on Build GRCh38

View Full Product Details