Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2209254-2209519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRICD5
Entrez Gene ID
283870
Gene Name
BRICHOS domain containing 5
Gene Aliases
C16orf79
UniGene
Hs.647399
Gene Chromosome Location
Chr.16: 2209253-2213037 on Build GRCh38
Transcripts
NM_182563.3 XM_005255263.4 NM_001199173.1 NM_001199174.1 NM_001199175.1 NM_022372.4 XM_005255475.3 XM_005255478.2 XM_005255479.2 XM_017023548.1 XM_017023549.1 XM_017023551.1

Gene Symbol
MLST8
Entrez Gene ID
64223
Gene Name
MTOR associated protein, LST8 homolog
Gene Aliases
GBL, GbetaL, LST8, POP3, WAT1
UniGene
Hs.29203
Gene Chromosome Location
Chr.16: 2205177-2209417 on Build GRCh38
Transcripts
NM_182563.3 XM_005255263.4 NM_001199173.1 NM_001199174.1 NM_001199175.1 NM_022372.4 XM_005255475.3 XM_005255478.2 XM_005255479.2 XM_017023548.1 XM_017023549.1 XM_017023551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5767564 c.755C>G p.S252W Chr.16: 2209390-2209390 on Build GRCh38
3999797 c.756G>T p.S252S Chr.16: 2209389-2209389 on Build GRCh38
5754822 c.695G>A p.R232Q Chr.16: 2209450-2209450 on Build GRCh38
5850474 c.755C>T p.S252L Chr.16: 2209390-2209390 on Build GRCh38
6025699 c.607C>T p.R203W Chr.16: 2209442-2209442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752577307 Chr.16: 2209349-2209349 on Build GRCh38
rs368529981 Chr.16: 2209339-2209339 on Build GRCh38
rs979604058 Chr.16: 2209312-2209312 on Build GRCh38
rs200527596 Chr.16: 2209405-2209405 on Build GRCh38
rs1056365976 Chr.16: 2209431-2209431 on Build GRCh38
rs564533621 Chr.16: 2209420-2209420 on Build GRCh38
rs368773570 Chr.16: 2209328-2209328 on Build GRCh38
rs140460866 Chr.16: 2209408-2209408 on Build GRCh38
rs554287445 Chr.16: 2209402-2209402 on Build GRCh38
rs766670397 Chr.16: 2209332-2209332 on Build GRCh38
rs371914671 Chr.16: 2209356-2209356 on Build GRCh38
rs192860278 Chr.16: 2209451-2209451 on Build GRCh38
rs939138515 Chr.16: 2209307-2209307 on Build GRCh38
rs923755913 Chr.16: 2209300-2209300 on Build GRCh38
rs758292937 Chr.16: 2209351-2209351 on Build GRCh38
rs753339753 Chr.16: 2209446-2209446 on Build GRCh38
rs755615527 Chr.16: 2209370-2209370 on Build GRCh38
rs747347986 Chr.16: 2209382-2209382 on Build GRCh38
rs142915435 Chr.16: 2209336-2209336 on Build GRCh38
rs771329821 Chr.16: 2209384-2209384 on Build GRCh38
rs753528058 Chr.16: 2209478-2209478 on Build GRCh38
rs200906793 Chr.16: 2209453-2209453 on Build GRCh38
rs774265809 Chr.16: 2209434-2209434 on Build GRCh38
rs766007844 Chr.16: 2209477-2209477 on Build GRCh38
rs760496856 Chr.16: 2209475-2209475 on Build GRCh38
rs773480492 Chr.16: 2209374-2209374 on Build GRCh38
rs372865097 Chr.16: 2209481-2209481 on Build GRCh38
rs199768716 Chr.16: 2209441-2209441 on Build GRCh38
rs370223216 Chr.16: 2209318-2209318 on Build GRCh38
rs200139247 Chr.16: 2209491-2209491 on Build GRCh38
rs776983175 Chr.16: 2209386-2209386 on Build GRCh38
rs201450400 Chr.16: 2209371-2209371 on Build GRCh38
rs762825593 Chr.16: 2209394-2209394 on Build GRCh38
rs758520564 Chr.16: 2209389-2209390 on Build GRCh38
rs750451769 Chr.16: 2209345-2209346 on Build GRCh38
rs780707788 Chr.16: 2209311-2209311 on Build GRCh38
rs200897869 Chr.16: 2209474-2209474 on Build GRCh38
rs372074545 Chr.16: 2209447-2209447 on Build GRCh38
rs745876382 Chr.16: 2209428-2209428 on Build GRCh38
rs879079800 Chr.16: 2209484-2209484 on Build GRCh38
rs199992378 Chr.16: 2209479-2209479 on Build GRCh38
rs976812717 Chr.16: 2209433-2209441 on Build GRCh38
rs745348677 Chr.16: 2209314-2209314 on Build GRCh38
rs771420086 Chr.16: 2209419-2209419 on Build GRCh38
rs756940448 Chr.16: 2209355-2209355 on Build GRCh38
rs772583788 Chr.16: 2209372-2209372 on Build GRCh38
rs763815040 Chr.16: 2209352-2209352 on Build GRCh38
rs757875029 Chr.16: 2209490-2209490 on Build GRCh38
rs758868939 Chr.16: 2209411-2209411 on Build GRCh38
rs765416492 Chr.16: 2209390-2209390 on Build GRCh38
rs376735983 Chr.16: 2209462-2209462 on Build GRCh38
rs1129185 Chr.16: 2209392-2209392 on Build GRCh38
rs111384111 Chr.16: 2209409-2209409 on Build GRCh38
rs766838136 Chr.16: 2209475-2209476 on Build GRCh38
rs940790069 Chr.16: 2209430-2209430 on Build GRCh38
rs773032940 Chr.16: 2209473-2209473 on Build GRCh38
rs561967169 Chr.16: 2209422-2209422 on Build GRCh38
rs367963964 Chr.16: 2209316-2209316 on Build GRCh38
rs138073931 Chr.16: 2209454-2209454 on Build GRCh38
rs372557457 Chr.16: 2209437-2209437 on Build GRCh38
rs898175158 Chr.16: 2209417-2209417 on Build GRCh38
rs766023464 Chr.16: 2209445-2209445 on Build GRCh38
rs779388829 Chr.16: 2209344-2209344 on Build GRCh38
rs769784762 Chr.16: 2209336-2209345 on Build GRCh38
rs372214827 Chr.16: 2209335-2209335 on Build GRCh38
rs761669699 Chr.16: 2209440-2209440 on Build GRCh38
rs773566730 Chr.16: 2209329-2209329 on Build GRCh38
rs751324990 Chr.16: 2209353-2209353 on Build GRCh38
rs1008313244 Chr.16: 2209373-2209373 on Build GRCh38
rs749392194 Chr.16: 2209457-2209457 on Build GRCh38
rs756494018 Chr.16: 2209493-2209493 on Build GRCh38
rs78285503 Chr.16: 2209389-2209389 on Build GRCh38
rs368994283 Chr.16: 2209464-2209464 on Build GRCh38
rs748611274 Chr.16: 2209326-2209326 on Build GRCh38
rs138333652 Chr.16: 2209414-2209414 on Build GRCh38
rs778322582 Chr.16: 2209412-2209412 on Build GRCh38
rs369383984 Chr.16: 2209463-2209463 on Build GRCh38
rs375593732 Chr.16: 2209401-2209401 on Build GRCh38
rs777961697 Chr.16: 2209407-2209407 on Build GRCh38
rs750873515 Chr.16: 2209492-2209492 on Build GRCh38
rs146244160 Chr.16: 2209442-2209442 on Build GRCh38
rs745438857 Chr.16: 2209358-2209358 on Build GRCh38
rs979183467 Chr.16: 2209391-2209391 on Build GRCh38
rs771219860 Chr.16: 2209424-2209424 on Build GRCh38
rs764057363 Chr.16: 2209395-2209395 on Build GRCh38
rs955557837 Chr.16: 2209360-2209360 on Build GRCh38
rs201717204 Chr.16: 2209450-2209450 on Build GRCh38
rs763064022 Chr.16: 2209425-2209425 on Build GRCh38
rs369257397 Chr.16: 2209362-2209362 on Build GRCh38
rs1042266228 Chr.16: 2209297-2209297 on Build GRCh38
rs767222604 Chr.16: 2209404-2209404 on Build GRCh38
rs142825627 Chr.16: 2209421-2209421 on Build GRCh38
rs1012103660 Chr.16: 2209432-2209432 on Build GRCh38
rs771956170 Chr.16: 2209472-2209472 on Build GRCh38
rs768530695 Chr.16: 2209427-2209427 on Build GRCh38

View Full Product Details