Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30651131-30651356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf136
Entrez Gene ID
221545
Gene Name
chromosome 6 open reading frame 136
Gene Aliases
-
UniGene
Hs.591787
Gene Chromosome Location
Chr.6: 30647039-30653210 on Build GRCh38
Transcripts
NM_001109938.2 NM_001161376.1 NM_145029.3 XM_011514384.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5648352 c.1197G>C p.L399L Chr.6: 30651356-30651356 on Build GRCh38
3684192 c.209T>G p.L70W Chr.6: 30651340-30651340 on Build GRCh38
6173723 c.205C>T p.R69C Chr.6: 30651336-30651336 on Build GRCh38
6616407 c.1178G>A p.R393H Chr.6: 30651337-30651337 on Build GRCh38
6616405 c.1144C>T p.R382C Chr.6: 30651303-30651303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766093375 Chr.6: 30651304-30651304 on Build GRCh38
rs763042784 Chr.6: 30651266-30651266 on Build GRCh38
rs1041681239 Chr.6: 30651169-30651169 on Build GRCh38
rs771419771 Chr.6: 30651221-30651221 on Build GRCh38
rs760527127 Chr.6: 30651297-30651297 on Build GRCh38
rs775598877 Chr.6: 30651250-30651250 on Build GRCh38
rs1040782014 Chr.6: 30651200-30651200 on Build GRCh38
rs747437491 Chr.6: 30651216-30651216 on Build GRCh38
rs575874165 Chr.6: 30651232-30651232 on Build GRCh38
rs769784783 Chr.6: 30651235-30651235 on Build GRCh38
rs1047035121 Chr.6: 30651165-30651165 on Build GRCh38
rs753798518 Chr.6: 30651323-30651323 on Build GRCh38
rs773122395 Chr.6: 30651179-30651179 on Build GRCh38
rs764422167 Chr.6: 30651281-30651281 on Build GRCh38
rs774539217 Chr.6: 30651288-30651288 on Build GRCh38
rs762911105 Chr.6: 30651271-30651271 on Build GRCh38
rs777283810 Chr.6: 30651229-30651229 on Build GRCh38
rs1005435842 Chr.6: 30651174-30651174 on Build GRCh38
rs373040690 Chr.6: 30651270-30651270 on Build GRCh38
rs141051300 Chr.6: 30651189-30651189 on Build GRCh38
rs770525226 Chr.6: 30651274-30651274 on Build GRCh38
rs554711801 Chr.6: 30651277-30651277 on Build GRCh38

View Full Product Details