Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75206431-75206656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf39
Entrez Gene ID
56905
Gene Name
chromosome 15 open reading frame 39
Gene Aliases
-
UniGene
Hs.17936
Gene Chromosome Location
Chr.15: 75201880-75212169 on Build GRCh38
Transcripts
NM_015492.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1198139 c.604G>T p.D202Y Chr.15: 75206652-75206652 on Build GRCh38
6195016 c.390C>G p.P130P Chr.15: 75206438-75206438 on Build GRCh38
5958343 c.453G>A p.V151V Chr.15: 75206501-75206501 on Build GRCh38
5198455 c.526C>T p.L176L Chr.15: 75206574-75206574 on Build GRCh38
1939035 c.577G>T p.G193W Chr.15: 75206625-75206625 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954201313 Chr.15: 75206516-75206516 on Build GRCh38
rs777024266 Chr.15: 75206465-75206465 on Build GRCh38
rs376379657 Chr.15: 75206527-75206527 on Build GRCh38
rs765148849 Chr.15: 75206551-75206551 on Build GRCh38
rs143072938 Chr.15: 75206533-75206533 on Build GRCh38
rs923165798 Chr.15: 75206529-75206529 on Build GRCh38
rs753578475 Chr.15: 75206534-75206534 on Build GRCh38
rs372183969 Chr.15: 75206552-75206552 on Build GRCh38
rs754575222 Chr.15: 75206541-75206541 on Build GRCh38
rs199696770 Chr.15: 75206523-75206523 on Build GRCh38
rs777874459 Chr.15: 75206571-75206571 on Build GRCh38
rs564716678 Chr.15: 75206633-75206633 on Build GRCh38
rs754035815 Chr.15: 75206452-75206452 on Build GRCh38
rs1048977016 Chr.15: 75206626-75206626 on Build GRCh38
rs910376894 Chr.15: 75206458-75206458 on Build GRCh38
rs371339122 Chr.15: 75206530-75206530 on Build GRCh38
rs866242565 Chr.15: 75206580-75206580 on Build GRCh38
rs746902353 Chr.15: 75206587-75206587 on Build GRCh38
rs749135337 Chr.15: 75206623-75206623 on Build GRCh38
rs151171215 Chr.15: 75206602-75206602 on Build GRCh38
rs374466401 Chr.15: 75206596-75206596 on Build GRCh38
rs746156594 Chr.15: 75206623-75206623 on Build GRCh38
rs143715170 Chr.15: 75206628-75206628 on Build GRCh38
rs747100347 Chr.15: 75206451-75206451 on Build GRCh38
rs777563887 Chr.15: 75206621-75206624 on Build GRCh38
rs548759069 Chr.15: 75206622-75206622 on Build GRCh38
rs770242057 Chr.15: 75206514-75206514 on Build GRCh38
rs746045130 Chr.15: 75206467-75206467 on Build GRCh38
rs749663017 Chr.15: 75206629-75206629 on Build GRCh38
rs758210261 Chr.15: 75206563-75206563 on Build GRCh38

View Full Product Details