Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114044885-114045134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADRB1
Entrez Gene ID
153
Gene Name
adrenoceptor beta 1
Gene Aliases
ADRB1R, B1AR, BETA1AR, RHR
UniGene
Hs.99913
Gene Chromosome Location
Chr.10: 114044047-114046908 on Build GRCh38
Transcripts
NM_000684.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4409515 c.822G>A p.S274S Chr.10: 114044954-114044954 on Build GRCh38
7301485 c.778A>C p.S260R Chr.10: 114044910-114044910 on Build GRCh38
4011736 c.811C>T p.R271W Chr.10: 114044943-114044943 on Build GRCh38
4011735 c.798C>T p.L266L Chr.10: 114044930-114044930 on Build GRCh38
5447989 c.928C>T p.R310W Chr.10: 114045060-114045060 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751522258 Chr.10: 114045043-114045043 on Build GRCh38
rs950307363 Chr.10: 114044969-114044969 on Build GRCh38
rs376897657 Chr.10: 114044954-114044954 on Build GRCh38
rs756619440 Chr.10: 114045109-114045109 on Build GRCh38
rs768923241 Chr.10: 114044941-114044941 on Build GRCh38
rs370424397 Chr.10: 114044955-114044955 on Build GRCh38
rs953727737 Chr.10: 114044917-114044917 on Build GRCh38
rs780900355 Chr.10: 114044931-114044931 on Build GRCh38
rs774798512 Chr.10: 114044945-114044945 on Build GRCh38
rs745308620 Chr.10: 114045062-114045062 on Build GRCh38
rs917506945 Chr.10: 114044960-114044960 on Build GRCh38
rs766569604 Chr.10: 114044959-114044959 on Build GRCh38
rs1027912819 Chr.10: 114044909-114044909 on Build GRCh38
rs238741 Chr.10: 114045084-114045084 on Build GRCh38
rs2773468 Chr.10: 114044936-114044936 on Build GRCh38
rs758176174 Chr.10: 114045010-114045010 on Build GRCh38
rs762200213 Chr.10: 114044946-114044947 on Build GRCh38
rs369955946 Chr.10: 114044911-114044911 on Build GRCh38
rs767559825 Chr.10: 114044953-114044953 on Build GRCh38
rs372579473 Chr.10: 114045060-114045060 on Build GRCh38
rs767675304 Chr.10: 114044947-114044952 on Build GRCh38
rs769031036 Chr.10: 114045078-114045078 on Build GRCh38
rs1048658242 Chr.10: 114044972-114045004 on Build GRCh38
rs750500075 Chr.10: 114044966-114044967 on Build GRCh38
rs780436351 Chr.10: 114044972-114044973 on Build GRCh38
rs936003506 Chr.10: 114045006-114045007 on Build GRCh38
rs754033628 Chr.10: 114044975-114044975 on Build GRCh38
rs777753862 Chr.10: 114045026-114045026 on Build GRCh38
rs772631299 Chr.10: 114045089-114045089 on Build GRCh38
rs779706805 Chr.10: 114045074-114045074 on Build GRCh38
rs622397 Chr.10: 114045103-114045103 on Build GRCh38
rs199698054 Chr.10: 114044940-114044940 on Build GRCh38
rs762344790 Chr.10: 114044951-114044951 on Build GRCh38
rs770536724 Chr.10: 114044914-114044914 on Build GRCh38
rs769199397 Chr.10: 114045070-114045070 on Build GRCh38
rs775010159 Chr.10: 114044935-114044935 on Build GRCh38
rs756167370 Chr.10: 114044970-114044970 on Build GRCh38
rs572172231 Chr.10: 114044994-114044994 on Build GRCh38
rs757322738 Chr.10: 114045048-114045048 on Build GRCh38
rs773726392 Chr.10: 114045110-114045110 on Build GRCh38

View Full Product Details