Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207126704-207126945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4BPA
Entrez Gene ID
722
Gene Name
complement component 4 binding protein alpha
Gene Aliases
C4BP, PRP
UniGene
Hs.1012
Gene Chromosome Location
Chr.1: 207104232-207144972 on Build GRCh38
Transcripts
NM_000715.3 XM_005273251.1 XM_005273252.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1338389 c.803delA p.G270fs*9 Chr.1: 207126809-207126809 on Build GRCh38
3400248 c.852T>C p.D284D Chr.1: 207126858-207126858 on Build GRCh38
463849 c.728A>C p.D243A Chr.1: 207126734-207126734 on Build GRCh38
414660 c.738T>A p.H246Q Chr.1: 207126744-207126744 on Build GRCh38
3482596 c.872C>T p.P291L Chr.1: 207126878-207126878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs997374620 Chr.1: 207126759-207126759 on Build GRCh38
rs767908699 Chr.1: 207126764-207126764 on Build GRCh38
rs573344831 Chr.1: 207126815-207126815 on Build GRCh38
rs201495889 Chr.1: 207126846-207126846 on Build GRCh38
rs775885891 Chr.1: 207126743-207126743 on Build GRCh38
rs750722809 Chr.1: 207126796-207126796 on Build GRCh38
rs777769501 Chr.1: 207126835-207126835 on Build GRCh38
rs921732366 Chr.1: 207126802-207126802 on Build GRCh38
rs778354731 Chr.1: 207126819-207126819 on Build GRCh38
rs774349261 Chr.1: 207126909-207126909 on Build GRCh38
rs202143924 Chr.1: 207126751-207126751 on Build GRCh38
rs746759203 Chr.1: 207126836-207126836 on Build GRCh38
rs376681896 Chr.1: 207126831-207126831 on Build GRCh38
rs530867822 Chr.1: 207126913-207126913 on Build GRCh38
rs2491393 Chr.1: 207126914-207126914 on Build GRCh38
rs146724239 Chr.1: 207126852-207126852 on Build GRCh38
rs761009386 Chr.1: 207126917-207126917 on Build GRCh38
rs1045706209 Chr.1: 207126895-207126895 on Build GRCh38
rs763077038 Chr.1: 207126750-207126750 on Build GRCh38
rs770073041 Chr.1: 207126742-207126742 on Build GRCh38
rs749558281 Chr.1: 207126866-207126866 on Build GRCh38
rs900584586 Chr.1: 207126729-207126729 on Build GRCh38
rs752953265 Chr.1: 207126877-207126877 on Build GRCh38
rs753541811 Chr.1: 207126807-207126807 on Build GRCh38
rs946084245 Chr.1: 207126885-207126885 on Build GRCh38
rs950393470 Chr.1: 207126739-207126739 on Build GRCh38
rs1010173219 Chr.1: 207126767-207126767 on Build GRCh38
rs1021922943 Chr.1: 207126826-207126826 on Build GRCh38
rs746812093 Chr.1: 207126730-207126730 on Build GRCh38
rs564131234 Chr.1: 207126921-207126921 on Build GRCh38
rs752667877 Chr.1: 207126825-207126825 on Build GRCh38
rs768773570 Chr.1: 207126894-207126894 on Build GRCh38
rs761062466 Chr.1: 207126797-207126797 on Build GRCh38
rs377164344 Chr.1: 207126893-207126893 on Build GRCh38
rs765970786 Chr.1: 207126799-207126799 on Build GRCh38
rs762291748 Chr.1: 207126762-207126762 on Build GRCh38
rs774887140 Chr.1: 207126760-207126760 on Build GRCh38

View Full Product Details