Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44666586-44666829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEIOC
Entrez Gene ID
284071
Gene Name
meiosis specific with coiled-coil domain
Gene Aliases
C17orf104
UniGene
Hs.349758
Gene Chromosome Location
Chr.17: 44656394-44677086 on Build GRCh38
Transcripts
NM_001145080.2 XM_005257236.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3191318 c.844G>C p.E282Q Chr.17: 44666755-44666755 on Build GRCh38
6434582 c.916C>A p.Q306K Chr.17: 44666827-44666827 on Build GRCh38
327593 c.824G>C p.G275A Chr.17: 44666735-44666735 on Build GRCh38
3989061 c.755C>T p.T252I Chr.17: 44666666-44666666 on Build GRCh38
5005416 c.722G>A p.R241Q Chr.17: 44666633-44666633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751799072 Chr.17: 44666701-44666701 on Build GRCh38
rs771621297 Chr.17: 44666774-44666774 on Build GRCh38
rs746194595 Chr.17: 44666780-44666780 on Build GRCh38
rs1052106703 Chr.17: 44666655-44666655 on Build GRCh38
rs752821525 Chr.17: 44666674-44666674 on Build GRCh38
rs532603101 Chr.17: 44666746-44666748 on Build GRCh38
rs1050263239 Chr.17: 44666768-44666768 on Build GRCh38
rs754561497 Chr.17: 44666754-44666754 on Build GRCh38
rs549436753 Chr.17: 44666678-44666678 on Build GRCh38
rs372645745 Chr.17: 44666767-44666767 on Build GRCh38
rs777112732 Chr.17: 44666779-44666779 on Build GRCh38
rs776796076 Chr.17: 44666619-44666619 on Build GRCh38
rs113545836 Chr.17: 44666688-44666688 on Build GRCh38
rs749202239 Chr.17: 44666748-44666748 on Build GRCh38
rs747807312 Chr.17: 44666769-44666769 on Build GRCh38
rs765646880 Chr.17: 44666671-44666671 on Build GRCh38
rs370251992 Chr.17: 44666751-44666751 on Build GRCh38
rs762988179 Chr.17: 44666684-44666684 on Build GRCh38
rs201395281 Chr.17: 44666704-44666704 on Build GRCh38
rs1022675033 Chr.17: 44666775-44666775 on Build GRCh38
rs538119359 Chr.17: 44666795-44666795 on Build GRCh38
rs866802792 Chr.17: 44666633-44666633 on Build GRCh38
rs781042706 Chr.17: 44666705-44666705 on Build GRCh38
rs369691539 Chr.17: 44666708-44666708 on Build GRCh38
rs868580957 Chr.17: 44666762-44666762 on Build GRCh38
rs909586149 Chr.17: 44666725-44666725 on Build GRCh38
rs756128513 Chr.17: 44666721-44666721 on Build GRCh38
rs779827818 Chr.17: 44666730-44666730 on Build GRCh38
rs760157460 Chr.17: 44666659-44666659 on Build GRCh38
rs992516091 Chr.17: 44666735-44666735 on Build GRCh38

View Full Product Details