Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57627609-57627840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALNT1
Entrez Gene ID
2583
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 1
Gene Aliases
GALGT, GALNACT, GalNAc-T, SPG26
UniGene
Hs.159481
Gene Chromosome Location
Chr.12: 57623409-57633239 on Build GRCh38
Transcripts
NM_001276468.1 NM_001478.4 XM_005268773.4 XM_011538147.2 XM_011538148.2 XM_017019140.1 XM_017019141.1 XM_017019142.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4575625 c.1190G>C p.R397P Chr.12: 57627812-57627812 on Build GRCh38
3667192 c.1188T>C p.Y396Y Chr.12: 57627814-57627814 on Build GRCh38
3676512 c.1221A>C p.P407P Chr.12: 57627781-57627781 on Build GRCh38
6609374 c.1324C>T p.R442W Chr.12: 57627678-57627678 on Build GRCh38
6609373 c.1300G>A p.G434S Chr.12: 57627702-57627702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374497472 Chr.12: 57627790-57627790 on Build GRCh38
rs752114167 Chr.12: 57627774-57627774 on Build GRCh38
rs377044701 Chr.12: 57627798-57627798 on Build GRCh38
rs746970658 Chr.12: 57627801-57627801 on Build GRCh38
rs768473421 Chr.12: 57627807-57627807 on Build GRCh38
rs893488082 Chr.12: 57627643-57627643 on Build GRCh38
rs763660125 Chr.12: 57627787-57627787 on Build GRCh38
rs555801359 Chr.12: 57627705-57627705 on Build GRCh38
rs763267291 Chr.12: 57627761-57627761 on Build GRCh38
rs865906473 Chr.12: 57627723-57627723 on Build GRCh38
rs749032499 Chr.12: 57627691-57627691 on Build GRCh38
rs762100777 Chr.12: 57627736-57627736 on Build GRCh38
rs750720880 Chr.12: 57627633-57627633 on Build GRCh38
rs143066025 Chr.12: 57627730-57627730 on Build GRCh38
rs755430965 Chr.12: 57627786-57627786 on Build GRCh38
rs779937342 Chr.12: 57627799-57627799 on Build GRCh38
rs563575730 Chr.12: 57627680-57627680 on Build GRCh38
rs756953267 Chr.12: 57627792-57627792 on Build GRCh38
rs752238904 Chr.12: 57627658-57627658 on Build GRCh38
rs766701585 Chr.12: 57627770-57627770 on Build GRCh38
rs371132455 Chr.12: 57627732-57627732 on Build GRCh38
rs745607251 Chr.12: 57627703-57627703 on Build GRCh38
rs1020205354 Chr.12: 57627762-57627762 on Build GRCh38
rs1052315467 Chr.12: 57627642-57627642 on Build GRCh38
rs777129843 Chr.12: 57627677-57627677 on Build GRCh38
rs1008645951 Chr.12: 57627751-57627751 on Build GRCh38
rs879255242 Chr.12: 57627704-57627704 on Build GRCh38
rs758922224 Chr.12: 57627645-57627645 on Build GRCh38
rs781389623 Chr.12: 57627812-57627812 on Build GRCh38
rs765320304 Chr.12: 57627752-57627752 on Build GRCh38
rs765701178 Chr.12: 57627631-57627631 on Build GRCh38
rs778860816 Chr.12: 57627700-57627700 on Build GRCh38
rs756790411 Chr.12: 57627697-57627697 on Build GRCh38
rs766825815 Chr.12: 57627647-57627647 on Build GRCh38
rs755693956 Chr.12: 57627670-57627670 on Build GRCh38

View Full Product Details