Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 109495215-109495488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK9
Entrez Gene ID
221264
Gene Name
adenylate kinase 9
Gene Aliases
AK 9, AKD1, AKD2, C6orf199, C6orf224, dJ70A9.1
UniGene
Hs.205144
Gene Chromosome Location
Chr.6: 109492855-109691253 on Build GRCh38
Transcripts
NM_001145128.2 XM_006715376.3 XM_011535550.2 XM_011535552.2 XM_011535553.2 XM_011535554.2 XM_011535558.1 XM_017010385.1 XM_017010387.1 XM_017010389.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
109825 c.556C>T p.P186S Chr.6: 109495397-109495397 on Build GRCh38
6470632 c.564A>C p.L188F Chr.6: 109495389-109495389 on Build GRCh38
6470634 c.563T>G p.L188* Chr.6: 109495390-109495390 on Build GRCh38
3828765 c.573G>A p.P191P Chr.6: 109495380-109495380 on Build GRCh38
3857396 c.5401C>T p.P1801S Chr.6: 109495355-109495355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780575487 Chr.6: 109495298-109495298 on Build GRCh38
rs571780605 Chr.6: 109495275-109495275 on Build GRCh38
rs769154274 Chr.6: 109495356-109495356 on Build GRCh38
rs144374788 Chr.6: 109495381-109495381 on Build GRCh38
rs1043948374 Chr.6: 109495261-109495261 on Build GRCh38
rs759240628 Chr.6: 109495401-109495401 on Build GRCh38
rs372963506 Chr.6: 109495433-109495433 on Build GRCh38
rs758694051 Chr.6: 109495459-109495459 on Build GRCh38
rs1021703379 Chr.6: 109495273-109495273 on Build GRCh38
rs369007124 Chr.6: 109495291-109495291 on Build GRCh38
rs201007213 Chr.6: 109495390-109495390 on Build GRCh38
rs772668381 Chr.6: 109495372-109495372 on Build GRCh38
rs373532544 Chr.6: 109495288-109495288 on Build GRCh38
rs747742013 Chr.6: 109495355-109495355 on Build GRCh38
rs776988724 Chr.6: 109495327-109495327 on Build GRCh38
rs750477059 Chr.6: 109495451-109495451 on Build GRCh38
rs1010232295 Chr.6: 109495258-109495258 on Build GRCh38
rs146609210 Chr.6: 109495437-109495437 on Build GRCh38
rs747307031 Chr.6: 109495309-109495309 on Build GRCh38
rs147982272 Chr.6: 109495397-109495397 on Build GRCh38
rs758649068 Chr.6: 109495294-109495294 on Build GRCh38
rs780416049 Chr.6: 109495460-109495460 on Build GRCh38
rs995209639 Chr.6: 109495406-109495406 on Build GRCh38
rs992196983 Chr.6: 109495452-109495452 on Build GRCh38
rs746127198 Chr.6: 109495293-109495293 on Build GRCh38
rs1007870340 Chr.6: 109495347-109495347 on Build GRCh38
rs760909547 Chr.6: 109495363-109495364 on Build GRCh38
rs767542340 Chr.6: 109495427-109495427 on Build GRCh38
rs1026816125 Chr.6: 109495419-109495419 on Build GRCh38
rs899620428 Chr.6: 109495391-109495391 on Build GRCh38
rs377494123 Chr.6: 109495289-109495289 on Build GRCh38
rs769270069 Chr.6: 109495320-109495320 on Build GRCh38
rs762615512 Chr.6: 109495373-109495373 on Build GRCh38
rs961012463 Chr.6: 109495450-109495450 on Build GRCh38
rs765433291 Chr.6: 109495440-109495440 on Build GRCh38
rs961699826 Chr.6: 109495443-109495443 on Build GRCh38
rs574248045 Chr.6: 109495384-109495384 on Build GRCh38
rs951178405 Chr.6: 109495424-109495424 on Build GRCh38

View Full Product Details