Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 75724729-75724970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADM
Entrez Gene ID
34
Gene Name
acyl-CoA dehydrogenase, C-4 to C-12 straight chain
Gene Aliases
ACAD1, MCAD, MCADH
UniGene
Hs.445040
Gene Chromosome Location
Chr.1: 75724347-75763679 on Build GRCh38
Transcripts
NM_000016.5 NM_001127328.2 NM_001286042.1 NM_001286043.1 NM_001286044.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5847310 c.26G>A p.C9Y Chr.1: 75724813-75724813 on Build GRCh38
32206 c.1A>G p.M1V Chr.1: 75724788-75724788 on Build GRCh38
5847311 c.26G>A p.C9Y Chr.1: 75724813-75724813 on Build GRCh38
5810274 c.18G>A p.G6G Chr.1: 75724805-75724805 on Build GRCh38
3492379 c.15C>T p.F5F Chr.1: 75724802-75724802 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949064040 Chr.1: 75724898-75724898 on Build GRCh38
rs201646499 Chr.1: 75724798-75724798 on Build GRCh38
rs1024056446 Chr.1: 75724802-75724802 on Build GRCh38
rs769935756 Chr.1: 75724812-75724812 on Build GRCh38
rs750745448 Chr.1: 75724857-75724857 on Build GRCh38
rs753756738 Chr.1: 75724778-75724778 on Build GRCh38
rs376904926 Chr.1: 75724764-75724764 on Build GRCh38
rs752190688 Chr.1: 75724793-75724793 on Build GRCh38
rs1047187888 Chr.1: 75724953-75724953 on Build GRCh38
rs753530746 Chr.1: 75724753-75724753 on Build GRCh38
rs757845443 Chr.1: 75724799-75724799 on Build GRCh38
rs369431746 Chr.1: 75724865-75724865 on Build GRCh38
rs906559903 Chr.1: 75724808-75724808 on Build GRCh38
rs1251075 Chr.1: 75724884-75724884 on Build GRCh38
rs545522153 Chr.1: 75724773-75724773 on Build GRCh38
rs772007972 Chr.1: 75724765-75724765 on Build GRCh38
rs995637184 Chr.1: 75724929-75724929 on Build GRCh38
rs751137059 Chr.1: 75724839-75724839 on Build GRCh38
rs768596219 Chr.1: 75724819-75724819 on Build GRCh38
rs770059186 Chr.1: 75724757-75724757 on Build GRCh38
rs749511161 Chr.1: 75724754-75724756 on Build GRCh38
rs781666604 Chr.1: 75724855-75724855 on Build GRCh38
rs759253844 Chr.1: 75724779-75724779 on Build GRCh38
rs758001595 Chr.1: 75724795-75724795 on Build GRCh38
rs774531501 Chr.1: 75724821-75724821 on Build GRCh38
rs760716158 Chr.1: 75724772-75724772 on Build GRCh38
rs764998071 Chr.1: 75724786-75724786 on Build GRCh38
rs116034618 Chr.1: 75724883-75724883 on Build GRCh38
rs763505014 Chr.1: 75724838-75724838 on Build GRCh38
rs1056022699 Chr.1: 75724923-75724923 on Build GRCh38
rs746244899 Chr.1: 75724809-75724809 on Build GRCh38
rs777389884 Chr.1: 75724797-75724797 on Build GRCh38
rs371787277 Chr.1: 75724750-75724750 on Build GRCh38
rs775488206 Chr.1: 75724758-75724758 on Build GRCh38
rs756493351 Chr.1: 75724860-75724860 on Build GRCh38
rs757646242 Chr.1: 75724847-75724847 on Build GRCh38
rs875989865 Chr.1: 75724782-75724793 on Build GRCh38
rs372088389 Chr.1: 75724804-75724804 on Build GRCh38
rs1057516778 Chr.1: 75724788-75724788 on Build GRCh38
rs59932454 Chr.1: 75724754-75724754 on Build GRCh38
rs367734665 Chr.1: 75724771-75724771 on Build GRCh38
rs762704029 Chr.1: 75724835-75724835 on Build GRCh38
rs763024539 Chr.1: 75724759-75724759 on Build GRCh38
rs953750279 Chr.1: 75724890-75724890 on Build GRCh38
rs760526492 Chr.1: 75724825-75724825 on Build GRCh38
rs746085082 Chr.1: 75724755-75724755 on Build GRCh38
rs759413479 Chr.1: 75724827-75724827 on Build GRCh38
rs369417973 Chr.1: 75724879-75724879 on Build GRCh38
rs529155733 Chr.1: 75724926-75724926 on Build GRCh38
rs764930767 Chr.1: 75724832-75724832 on Build GRCh38
rs747346853 Chr.1: 75724751-75724751 on Build GRCh38
rs937619461 Chr.1: 75724916-75724916 on Build GRCh38
rs1046526722 Chr.1: 75724906-75724906 on Build GRCh38
rs561520228 Chr.1: 75724920-75724920 on Build GRCh38
rs749483210 Chr.1: 75724867-75724867 on Build GRCh38
rs749318140 Chr.1: 75724815-75724815 on Build GRCh38
rs375492495 Chr.1: 75724828-75724828 on Build GRCh38
rs540887454 Chr.1: 75724944-75724944 on Build GRCh38
rs926291389 Chr.1: 75724912-75724912 on Build GRCh38
rs770961707 Chr.1: 75724826-75724826 on Build GRCh38
rs771491707 Chr.1: 75724765-75724765 on Build GRCh38
rs780284821 Chr.1: 75724813-75724813 on Build GRCh38

View Full Product Details