Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 32599769-32600032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD27
Entrez Gene ID
84079
Gene Name
ankyrin repeat domain 27
Gene Aliases
PP12899, VARP
UniGene
Hs.59236
Gene Chromosome Location
Chr.19: 32597001-32675215 on Build GRCh38
Transcripts
NM_032139.2 XM_006723412.2 XM_017027349.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4076721 c.2820C>T p.Y940Y Chr.19: 32599998-32599998 on Build GRCh38
7430445 c.2814G>C p.Q938H Chr.19: 32600004-32600004 on Build GRCh38
6242568 c.2846A>G p.K949R Chr.19: 32599972-32599972 on Build GRCh38
5071948 c.2768-260_2846+71del410 p.? Chr.19: 32599901-32600310 on Build GRCh38
439205 c.2790A>G p.L930L Chr.19: 32600028-32600028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050346895 Chr.19: 32599841-32599841 on Build GRCh38
rs774787728 Chr.19: 32600000-32600000 on Build GRCh38
rs763353897 Chr.19: 32599970-32599973 on Build GRCh38
rs749189618 Chr.19: 32599931-32599931 on Build GRCh38
rs556253587 Chr.19: 32599864-32599864 on Build GRCh38
rs889135258 Chr.19: 32599844-32599844 on Build GRCh38
rs370412682 Chr.19: 32599803-32599803 on Build GRCh38
rs1047785115 Chr.19: 32599843-32599843 on Build GRCh38
rs899286109 Chr.19: 32599815-32599815 on Build GRCh38
rs988045182 Chr.19: 32599970-32599970 on Build GRCh38
rs776983334 Chr.19: 32599953-32599953 on Build GRCh38
rs780012231 Chr.19: 32599930-32599930 on Build GRCh38
rs760027460 Chr.19: 32599957-32599957 on Build GRCh38
rs775959510 Chr.19: 32599966-32599966 on Build GRCh38
rs746400251 Chr.19: 32599916-32599916 on Build GRCh38
rs970388032 Chr.19: 32599869-32599869 on Build GRCh38
rs79952649 Chr.19: 32599933-32599933 on Build GRCh38
rs756946700 Chr.19: 32599806-32599806 on Build GRCh38
rs1000106762 Chr.19: 32599872-32599872 on Build GRCh38
rs769271972 Chr.19: 32599971-32599972 on Build GRCh38
rs758742646 Chr.19: 32599983-32599983 on Build GRCh38
rs892611530 Chr.19: 32599894-32599894 on Build GRCh38
rs576130475 Chr.19: 32599868-32599868 on Build GRCh38
rs754949637 Chr.19: 32599998-32599998 on Build GRCh38
rs763692565 Chr.19: 32599797-32599797 on Build GRCh38
rs751083566 Chr.19: 32599980-32599980 on Build GRCh38
rs746995939 Chr.19: 32599941-32599941 on Build GRCh38
rs749382232 Chr.19: 32599862-32599864 on Build GRCh38
rs368345607 Chr.19: 32599936-32599937 on Build GRCh38
rs779354273 Chr.19: 32599900-32599900 on Build GRCh38
rs750051043 Chr.19: 32599985-32599985 on Build GRCh38
rs1009779396 Chr.19: 32599895-32599895 on Build GRCh38
rs780641640 Chr.19: 32599816-32599816 on Build GRCh38
rs955674085 Chr.19: 32599898-32599898 on Build GRCh38
rs79274059 Chr.19: 32599851-32599851 on Build GRCh38
rs368592113 Chr.19: 32599944-32599944 on Build GRCh38
rs763577757 Chr.19: 32599969-32599969 on Build GRCh38
rs779852914 Chr.19: 32599989-32599989 on Build GRCh38
rs372391792 Chr.19: 32599987-32599987 on Build GRCh38
rs1022458568 Chr.19: 32599911-32599911 on Build GRCh38
rs749028727 Chr.19: 32599992-32599992 on Build GRCh38
rs967788335 Chr.19: 32599912-32599912 on Build GRCh38
rs755886166 Chr.19: 32599828-32599828 on Build GRCh38
rs767254378 Chr.19: 32599984-32599984 on Build GRCh38
rs764449382 Chr.19: 32599974-32599974 on Build GRCh38
rs1032719152 Chr.19: 32599873-32599873 on Build GRCh38
rs780954059 Chr.19: 32599909-32599910 on Build GRCh38

View Full Product Details