Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64895198-64895467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG2A
Entrez Gene ID
23130
Gene Name
autophagy related 2A
Gene Aliases
-
UniGene
Hs.370671
Gene Chromosome Location
Chr.11: 64894532-64917250 on Build GRCh38
Transcripts
NM_015104.2 XM_005273849.3 XM_005273850.3 XM_011544863.2 XM_011544864.2 XM_011544865.2 XM_011544866.2 XM_011544868.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1298396 c.5459C>A p.S1820Y Chr.11: 64895411-64895411 on Build GRCh38
6606646 c.5575C>T p.R1859* Chr.11: 64895295-64895295 on Build GRCh38
7230943 c.5562C>G p.A1854A Chr.11: 64895308-64895308 on Build GRCh38
4146082 c.5493T>C p.D1831D Chr.11: 64895377-64895377 on Build GRCh38
3451624 c.5484C>T p.S1828S Chr.11: 64895386-64895386 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754445707 Chr.11: 64895233-64895233 on Build GRCh38
rs375146931 Chr.11: 64895428-64895428 on Build GRCh38
rs373838276 Chr.11: 64895304-64895304 on Build GRCh38
rs749347513 Chr.11: 64895404-64895404 on Build GRCh38
rs777430718 Chr.11: 64895243-64895243 on Build GRCh38
rs746753700 Chr.11: 64895247-64895247 on Build GRCh38
rs765947965 Chr.11: 64895232-64895232 on Build GRCh38
rs763410775 Chr.11: 64895218-64895218 on Build GRCh38
rs759188147 Chr.11: 64895370-64895370 on Build GRCh38
rs140343176 Chr.11: 64895378-64895378 on Build GRCh38
rs764339266 Chr.11: 64895224-64895224 on Build GRCh38
rs371652521 Chr.11: 64895295-64895295 on Build GRCh38
rs776942106 Chr.11: 64895265-64895265 on Build GRCh38
rs547893861 Chr.11: 64895235-64895235 on Build GRCh38
rs371145155 Chr.11: 64895354-64895354 on Build GRCh38
rs181554393 Chr.11: 64895335-64895335 on Build GRCh38
rs772492714 Chr.11: 64895413-64895413 on Build GRCh38
rs536644755 Chr.11: 64895332-64895332 on Build GRCh38
rs935545659 Chr.11: 64895348-64895348 on Build GRCh38
rs142328414 Chr.11: 64895362-64895362 on Build GRCh38
rs762178399 Chr.11: 64895287-64895287 on Build GRCh38
rs370496305 Chr.11: 64895366-64895366 on Build GRCh38
rs535616947 Chr.11: 64895391-64895391 on Build GRCh38
rs757034647 Chr.11: 64895352-64895352 on Build GRCh38
rs765819718 Chr.11: 64895239-64895242 on Build GRCh38
rs779340270 Chr.11: 64895408-64895408 on Build GRCh38
rs773575172 Chr.11: 64895419-64895419 on Build GRCh38
rs148750841 Chr.11: 64895355-64895355 on Build GRCh38
rs146530015 Chr.11: 64895427-64895427 on Build GRCh38
rs201688296 Chr.11: 64895305-64895305 on Build GRCh38
rs373093396 Chr.11: 64895246-64895246 on Build GRCh38
rs774210181 Chr.11: 64895447-64895447 on Build GRCh38
rs374095968 Chr.11: 64895385-64895385 on Build GRCh38
rs1027862660 Chr.11: 64895372-64895372 on Build GRCh38
rs866299313 Chr.11: 64895423-64895423 on Build GRCh38
rs1044101332 Chr.11: 64895360-64895360 on Build GRCh38
rs755716969 Chr.11: 64895231-64895231 on Build GRCh38
rs751480306 Chr.11: 64895388-64895388 on Build GRCh38
rs200184532 Chr.11: 64895363-64895363 on Build GRCh38
rs745973761 Chr.11: 64895268-64895268 on Build GRCh38
rs77802938 Chr.11: 64895377-64895377 on Build GRCh38
rs200987274 Chr.11: 64895230-64895230 on Build GRCh38
rs771075448 Chr.11: 64895260-64895260 on Build GRCh38
rs588060 Chr.11: 64895274-64895274 on Build GRCh38
rs759096270 Chr.11: 64895298-64895298 on Build GRCh38
rs747321490 Chr.11: 64895421-64895421 on Build GRCh38
rs776184900 Chr.11: 64895273-64895273 on Build GRCh38
rs748675061 Chr.11: 64895411-64895411 on Build GRCh38
rs757163377 Chr.11: 64895390-64895390 on Build GRCh38
rs769461754 Chr.11: 64895426-64895426 on Build GRCh38
rs377418719 Chr.11: 64895258-64895258 on Build GRCh38
rs769131121 Chr.11: 64895276-64895276 on Build GRCh38
rs780263618 Chr.11: 64895402-64895402 on Build GRCh38
rs774581151 Chr.11: 64895281-64895281 on Build GRCh38
rs763640027 Chr.11: 64895329-64895329 on Build GRCh38
rs937396540 Chr.11: 64895410-64895410 on Build GRCh38
rs369212911 Chr.11: 64895240-64895240 on Build GRCh38
rs369082773 Chr.11: 64895275-64895275 on Build GRCh38
rs768751201 Chr.11: 64895407-64895407 on Build GRCh38
rs766052424 Chr.11: 64895294-64895294 on Build GRCh38
rs144231884 Chr.11: 64895353-64895353 on Build GRCh38
rs756409677 Chr.11: 64895398-64895398 on Build GRCh38
rs778862990 Chr.11: 64895234-64895234 on Build GRCh38
rs1056716173 Chr.11: 64895351-64895351 on Build GRCh38
rs762126437 Chr.11: 64895364-64895364 on Build GRCh38
rs751337435 Chr.11: 64895347-64895347 on Build GRCh38
rs148192310 Chr.11: 64895397-64895397 on Build GRCh38
rs375890649 Chr.11: 64895365-64895365 on Build GRCh38
rs769959423 Chr.11: 64895271-64895271 on Build GRCh38
rs763633419 Chr.11: 64895431-64895431 on Build GRCh38
rs762478986 Chr.11: 64895383-64895383 on Build GRCh38

View Full Product Details