Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181055220-181055493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL9
Entrez Gene ID
153579
Gene Name
butyrophilin like 9
Gene Aliases
BTN3, BTN8, VDLS1900
UniGene
Hs.546502
Gene Chromosome Location
Chr.5: 181039875-181061525 on Build GRCh38
Transcripts
NM_001308245.1 NM_152547.4 XM_006714823.3 XM_006714824.3 XM_006714825.3 XM_011534444.2 XM_017009113.1 XM_017009114.1 XM_017009115.1 XM_017009116.1 XM_017009117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4512797 c.916C>T p.Q306* Chr.5: 181055441-181055441 on Build GRCh38
4512798 c.916C>T p.Q306* Chr.5: 181055441-181055441 on Build GRCh38
7357872 c.912G>T p.K304N Chr.5: 181055437-181055437 on Build GRCh38
7357873 c.912G>T p.K304N Chr.5: 181055437-181055437 on Build GRCh38
5823169 c.908-1G>T p.? Chr.5: 181055432-181055432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188417375 Chr.5: 181055269-181055269 on Build GRCh38
rs761241919 Chr.5: 181055418-181055418 on Build GRCh38
rs10479601 Chr.5: 181055331-181055331 on Build GRCh38
rs773913520 Chr.5: 181055460-181055461 on Build GRCh38
rs774457661 Chr.5: 181055474-181055474 on Build GRCh38
rs748742004 Chr.5: 181055419-181055419 on Build GRCh38
rs748471488 Chr.5: 181055463-181055463 on Build GRCh38
rs769074522 Chr.5: 181055453-181055453 on Build GRCh38
rs143224692 Chr.5: 181055437-181055437 on Build GRCh38
rs768608671 Chr.5: 181055430-181055430 on Build GRCh38
rs368314229 Chr.5: 181055324-181055324 on Build GRCh38
rs1043575245 Chr.5: 181055462-181055462 on Build GRCh38
rs768383106 Chr.5: 181055415-181055418 on Build GRCh38
rs777582106 Chr.5: 181055392-181055392 on Build GRCh38
rs770978988 Chr.5: 181055473-181055473 on Build GRCh38
rs11951358 Chr.5: 181055250-181055250 on Build GRCh38
rs773416054 Chr.5: 181055467-181055467 on Build GRCh38
rs879217494 Chr.5: 181055317-181055317 on Build GRCh38
rs747517924 Chr.5: 181055440-181055440 on Build GRCh38
rs769988503 Chr.5: 181055464-181055464 on Build GRCh38
rs781571300 Chr.5: 181055454-181055454 on Build GRCh38
rs878954720 Chr.5: 181055304-181055304 on Build GRCh38
rs756824735 Chr.5: 181055426-181055426 on Build GRCh38
rs752470872 Chr.5: 181055388-181055388 on Build GRCh38
rs915840793 Chr.5: 181055370-181055370 on Build GRCh38
rs145688786 Chr.5: 181055390-181055390 on Build GRCh38
rs112262574 Chr.5: 181055303-181055304 on Build GRCh38
rs200210432 Chr.5: 181055377-181055377 on Build GRCh38
rs184688226 Chr.5: 181055469-181055469 on Build GRCh38

View Full Product Details