Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1244378-1244651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5D
Entrez Gene ID
513
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, delta subunit
Gene Aliases
-
UniGene
Hs.418668
Gene Chromosome Location
Chr.19: 1241750-1244825 on Build GRCh38
Transcripts
NM_001001975.1 NM_001687.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6049475 c.452C>T p.A151V Chr.19: 1244382-1244382 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs140196501 Chr.19: 1244422-1244422 on Build GRCh38
rs184673680 Chr.19: 1244482-1244482 on Build GRCh38
rs747579588 Chr.19: 1244445-1244445 on Build GRCh38
rs555922603 Chr.19: 1244615-1244615 on Build GRCh38
rs542366555 Chr.19: 1244601-1244601 on Build GRCh38
rs774150264 Chr.19: 1244455-1244455 on Build GRCh38
rs779296438 Chr.19: 1244431-1244431 on Build GRCh38
rs735524 Chr.19: 1244519-1244519 on Build GRCh38
rs534386038 Chr.19: 1244450-1244450 on Build GRCh38
rs977710081 Chr.19: 1244525-1244525 on Build GRCh38
rs957577698 Chr.19: 1244548-1244548 on Build GRCh38
rs750661675 Chr.19: 1244439-1244439 on Build GRCh38
rs200593395 Chr.19: 1244402-1244402 on Build GRCh38
rs1063633 Chr.19: 1244611-1244611 on Build GRCh38
rs1022744480 Chr.19: 1244476-1244476 on Build GRCh38
rs370466468 Chr.19: 1244411-1244411 on Build GRCh38
rs916740784 Chr.19: 1244564-1244564 on Build GRCh38
rs978058175 Chr.19: 1244486-1244486 on Build GRCh38
rs760754622 Chr.19: 1244477-1244477 on Build GRCh38
rs372935723 Chr.19: 1244404-1244404 on Build GRCh38
rs780505538 Chr.19: 1244444-1244444 on Build GRCh38
rs758810002 Chr.19: 1244440-1244440 on Build GRCh38
rs991428422 Chr.19: 1244557-1244557 on Build GRCh38
rs377518580 Chr.19: 1244458-1244458 on Build GRCh38
rs908088829 Chr.19: 1244628-1244628 on Build GRCh38
rs981913586 Chr.19: 1244594-1244594 on Build GRCh38
rs772607438 Chr.19: 1244451-1244451 on Build GRCh38
rs1007694553 Chr.19: 1244488-1244488 on Build GRCh38
rs762573715 Chr.19: 1244397-1244397 on Build GRCh38
rs375864876 Chr.19: 1244410-1244410 on Build GRCh38
rs767462384 Chr.19: 1244459-1244459 on Build GRCh38
rs865966395 Chr.19: 1244417-1244417 on Build GRCh38
rs769336953 Chr.19: 1244396-1244396 on Build GRCh38
rs764213151 Chr.19: 1244481-1244481 on Build GRCh38
rs764335748 Chr.19: 1244425-1244425 on Build GRCh38
rs960742879 Chr.19: 1244537-1244537 on Build GRCh38
rs1040110515 Chr.19: 1244432-1244432 on Build GRCh38
rs373551178 Chr.19: 1244426-1244426 on Build GRCh38
rs755565492 Chr.19: 1244593-1244593 on Build GRCh38
rs948859084 Chr.19: 1244585-1244585 on Build GRCh38
rs200093223 Chr.19: 1244453-1244453 on Build GRCh38
rs766143982 Chr.19: 1244399-1244399 on Build GRCh38
rs1036120578 Chr.19: 1244620-1244620 on Build GRCh38
rs571314147 Chr.19: 1244474-1244474 on Build GRCh38
rs112343618 Chr.19: 1244441-1244441 on Build GRCh38
rs372801460 Chr.19: 1244409-1244409 on Build GRCh38
rs770525531 Chr.19: 1244454-1244454 on Build GRCh38
rs915867444 Chr.19: 1244570-1244570 on Build GRCh38
rs774017721 Chr.19: 1244401-1244401 on Build GRCh38
rs114270333 Chr.19: 1244608-1244608 on Build GRCh38

View Full Product Details