Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119229890-119230153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT4
Entrez Gene ID
8702
Gene Name
beta-1,4-galactosyltransferase 4
Gene Aliases
B4Gal-T4, beta4Gal-T4
UniGene
Hs.13225
Gene Chromosome Location
Chr.3: 119211742-119240946 on Build GRCh38
Transcripts
NM_003778.3 NM_212543.1 XM_005247855.1 XM_006713798.2 XM_006713799.2 XM_006713800.1 XM_006713801.2 XM_011513260.1 XM_017007401.1 NR_046574.1

Gene Symbol
B4GALT4-AS1
Entrez Gene ID
100874201
Gene Name
B4GALT4 antisense RNA 1
Gene Aliases
-
UniGene
Hs.721307
Gene Chromosome Location
Chr.3: 119226486-119235141 on Build GRCh38
Transcripts
NM_003778.3 NM_212543.1 XM_005247855.1 XM_006713798.2 XM_006713799.2 XM_006713800.1 XM_006713801.2 XM_011513260.1 XM_017007401.1 NR_046574.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4112507 c.191C>T p.T64I Chr.3: 119229909-119229909 on Build GRCh38
3585725 c.115C>T p.Q39* Chr.3: 119229985-119229985 on Build GRCh38
4112506 c.206C>T p.T69M Chr.3: 119229894-119229894 on Build GRCh38
7147998 c.159G>A p.K53K Chr.3: 119229941-119229941 on Build GRCh38
7125712 c.115C>G p.Q39E Chr.3: 119229985-119229985 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745487394 Chr.3: 119229947-119229947 on Build GRCh38
rs758939498 Chr.3: 119229916-119229916 on Build GRCh38
rs368354053 Chr.3: 119230112-119230112 on Build GRCh38
rs757825010 Chr.3: 119230084-119230084 on Build GRCh38
rs757589219 Chr.3: 119229938-119229938 on Build GRCh38
rs146652873 Chr.3: 119230119-119230119 on Build GRCh38
rs764616640 Chr.3: 119230071-119230071 on Build GRCh38
rs910569200 Chr.3: 119229920-119229920 on Build GRCh38
rs750873559 Chr.3: 119230111-119230111 on Build GRCh38
rs543706671 Chr.3: 119230011-119230011 on Build GRCh38
rs915357416 Chr.3: 119229969-119229969 on Build GRCh38
rs72655931 Chr.3: 119230131-119230131 on Build GRCh38
rs898744501 Chr.3: 119230052-119230052 on Build GRCh38
rs764357413 Chr.3: 119230019-119230019 on Build GRCh38
rs778148279 Chr.3: 119229924-119229924 on Build GRCh38
rs773041353 Chr.3: 119230007-119230007 on Build GRCh38
rs774168959 Chr.3: 119229991-119229991 on Build GRCh38
rs772205965 Chr.3: 119230059-119230059 on Build GRCh38
rs34746912 Chr.3: 119229998-119229998 on Build GRCh38
rs748773067 Chr.3: 119230114-119230114 on Build GRCh38
rs1053768969 Chr.3: 119229959-119229959 on Build GRCh38
rs560745852 Chr.3: 119229951-119229951 on Build GRCh38
rs768073001 Chr.3: 119230117-119230117 on Build GRCh38
rs772275586 Chr.3: 119229971-119229971 on Build GRCh38
rs772056290 Chr.3: 119230125-119230125 on Build GRCh38
rs759742274 Chr.3: 119230008-119230008 on Build GRCh38
rs781671415 Chr.3: 119230088-119230088 on Build GRCh38
rs72655932 Chr.3: 119230070-119230070 on Build GRCh38
rs748870417 Chr.3: 119229972-119229972 on Build GRCh38
rs771156985 Chr.3: 119230124-119230124 on Build GRCh38
rs1055273361 Chr.3: 119230006-119230006 on Build GRCh38
rs752039854 Chr.3: 119230082-119230082 on Build GRCh38
rs564527519 Chr.3: 119230012-119230012 on Build GRCh38
rs747344665 Chr.3: 119229925-119229925 on Build GRCh38
rs72655933 Chr.3: 119229983-119229983 on Build GRCh38
rs568412801 Chr.3: 119230126-119230126 on Build GRCh38

View Full Product Details