Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97314552-97314779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC180
Entrez Gene ID
100499483
Gene Name
coiled-coil domain containing 180
Gene Aliases
BDAG1, C9orf174, KIAA1529
UniGene
Hs.435629
Gene Chromosome Location
Chr.9: 97307628-97377295 on Build GRCh38
Transcripts
NM_020893.2 NR_036527.1 NR_036528.1 NR_036529.1

Gene Symbol
LOC100499484-C9ORF174
Entrez Gene ID
57653
Gene Name
LOC100499484-C9orf174 readthrough
Gene Aliases
-
UniGene
Hs.435629
Gene Chromosome Location
Chr.9: 97238426-97377295 on Build GRCh38
Transcripts
NM_020893.2 NR_036527.1 NR_036528.1 NR_036529.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1624334 c.767G>A p.R256Q Chr.9: 97314664-97314664 on Build GRCh38
5759262 c.763C>T p.Q255* Chr.9: 97314669-97314669 on Build GRCh38
4559979 c.817G>A p.D273N Chr.9: 97314723-97314723 on Build GRCh38
5759261 c.1189C>T p.Q397* Chr.9: 97314669-97314669 on Build GRCh38
6621912 c.1191G>T p.Q397H Chr.9: 97314671-97314671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145582319 Chr.9: 97314648-97314648 on Build GRCh38
rs756264154 Chr.9: 97314643-97314657 on Build GRCh38
rs375737210 Chr.9: 97314753-97314753 on Build GRCh38
rs745660096 Chr.9: 97314714-97314714 on Build GRCh38
rs758771457 Chr.9: 97314584-97314584 on Build GRCh38
rs377007527 Chr.9: 97314652-97314652 on Build GRCh38
rs1013597089 Chr.9: 97314587-97314587 on Build GRCh38
rs765412824 Chr.9: 97314663-97314663 on Build GRCh38
rs770537232 Chr.9: 97314709-97314709 on Build GRCh38
rs755351156 Chr.9: 97314695-97314695 on Build GRCh38
rs3747502 Chr.9: 97314614-97314614 on Build GRCh38
rs766688793 Chr.9: 97314735-97314735 on Build GRCh38
rs769241328 Chr.9: 97314610-97314610 on Build GRCh38
rs774039301 Chr.9: 97314713-97314713 on Build GRCh38
rs201480101 Chr.9: 97314718-97314718 on Build GRCh38
rs780526633 Chr.9: 97314665-97314665 on Build GRCh38
rs748673740 Chr.9: 97314703-97314703 on Build GRCh38
rs745726441 Chr.9: 97314634-97314634 on Build GRCh38
rs771982374 Chr.9: 97314717-97314717 on Build GRCh38
rs751791769 Chr.9: 97314736-97314736 on Build GRCh38
rs763168905 Chr.9: 97314725-97314725 on Build GRCh38
rs774159388 Chr.9: 97314651-97314651 on Build GRCh38
rs376687587 Chr.9: 97314585-97314585 on Build GRCh38
rs199694770 Chr.9: 97314647-97314647 on Build GRCh38
rs769248304 Chr.9: 97314625-97314625 on Build GRCh38
rs140321791 Chr.9: 97314722-97314722 on Build GRCh38
rs781716815 Chr.9: 97314699-97314699 on Build GRCh38
rs758716537 Chr.9: 97314664-97314664 on Build GRCh38
rs761946201 Chr.9: 97314661-97314661 on Build GRCh38
rs748198969 Chr.9: 97314742-97314742 on Build GRCh38
rs772780982 Chr.9: 97314630-97314630 on Build GRCh38
rs986523565 Chr.9: 97314740-97314740 on Build GRCh38
rs145581112 Chr.9: 97314723-97314723 on Build GRCh38
rs747470456 Chr.9: 97314604-97314604 on Build GRCh38
rs752632555 Chr.9: 97314654-97314654 on Build GRCh38
rs955139881 Chr.9: 97314586-97314586 on Build GRCh38
rs1032532866 Chr.9: 97314681-97314681 on Build GRCh38
rs373860685 Chr.9: 97314704-97314704 on Build GRCh38
rs372488258 Chr.9: 97314752-97314752 on Build GRCh38
rs751945259 Chr.9: 97314687-97314687 on Build GRCh38

View Full Product Details