Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57235706-57235979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APLNR
Entrez Gene ID
187
Gene Name
apelin receptor
Gene Aliases
AGTRL1, APJ, APJR, HG11
UniGene
Hs.438311
Gene Chromosome Location
Chr.11: 57233577-57237453 on Build GRCh38
Transcripts
NM_005161.4 NR_027991.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1704006 c.1111C>T p.P371S Chr.11: 57235894-57235894 on Build GRCh38
96302 c.1099G>A p.E367K Chr.11: 57235906-57235906 on Build GRCh38
3953417 c.1085G>T p.G362V Chr.11: 57235920-57235920 on Build GRCh38
5372798 c.1067C>T p.P356L Chr.11: 57235938-57235938 on Build GRCh38
7290289 c.1064G>A p.G355D Chr.11: 57235941-57235941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371814515 Chr.11: 57235833-57235833 on Build GRCh38
rs759461727 Chr.11: 57235889-57235889 on Build GRCh38
rs773385120 Chr.11: 57235855-57235855 on Build GRCh38
rs1048402746 Chr.11: 57235939-57235939 on Build GRCh38
rs987154596 Chr.11: 57235942-57235942 on Build GRCh38
rs959994604 Chr.11: 57235835-57235835 on Build GRCh38
rs142294759 Chr.11: 57235907-57235907 on Build GRCh38
rs753489933 Chr.11: 57235924-57235924 on Build GRCh38
rs776757052 Chr.11: 57235842-57235842 on Build GRCh38
rs767481092 Chr.11: 57235906-57235906 on Build GRCh38
rs754256216 Chr.11: 57235828-57235829 on Build GRCh38
rs147773675 Chr.11: 57235946-57235946 on Build GRCh38
rs751341256 Chr.11: 57235874-57235874 on Build GRCh38
rs748258607 Chr.11: 57235952-57235952 on Build GRCh38
rs148940731 Chr.11: 57235810-57235810 on Build GRCh38
rs558188721 Chr.11: 57235734-57235734 on Build GRCh38
rs777596520 Chr.11: 57235915-57235915 on Build GRCh38
rs755228199 Chr.11: 57235842-57235843 on Build GRCh38
rs761824680 Chr.11: 57235845-57235845 on Build GRCh38
rs201061974 Chr.11: 57235826-57235826 on Build GRCh38
rs544882959 Chr.11: 57235831-57235831 on Build GRCh38
rs751405218 Chr.11: 57235820-57235820 on Build GRCh38
rs376524097 Chr.11: 57235943-57235943 on Build GRCh38
rs780320730 Chr.11: 57235934-57235934 on Build GRCh38
rs762760753 Chr.11: 57235859-57235859 on Build GRCh38
rs780631771 Chr.11: 57235834-57235834 on Build GRCh38
rs200674887 Chr.11: 57235837-57235837 on Build GRCh38
rs779844962 Chr.11: 57235791-57235791 on Build GRCh38
rs146062493 Chr.11: 57235926-57235926 on Build GRCh38
rs749069536 Chr.11: 57235838-57235838 on Build GRCh38
rs766139270 Chr.11: 57235869-57235869 on Build GRCh38
rs376606919 Chr.11: 57235821-57235821 on Build GRCh38
rs781612967 Chr.11: 57235951-57235951 on Build GRCh38
rs566944394 Chr.11: 57235783-57235783 on Build GRCh38
rs375912159 Chr.11: 57235885-57235885 on Build GRCh38
rs755891901 Chr.11: 57235830-57235830 on Build GRCh38
rs899085863 Chr.11: 57235733-57235733 on Build GRCh38
rs995076303 Chr.11: 57235758-57235758 on Build GRCh38
rs755614745 Chr.11: 57235909-57235909 on Build GRCh38
rs770009487 Chr.11: 57235848-57235848 on Build GRCh38
rs534306738 Chr.11: 57235796-57235796 on Build GRCh38
rs147023435 Chr.11: 57235836-57235836 on Build GRCh38
rs977795947 Chr.11: 57235846-57235846 on Build GRCh38

View Full Product Details