Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 129605768-129606017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP18
Entrez Gene ID
93663
Gene Name
Rho GTPase activating protein 18
Gene Aliases
MacGAP, SENEX, bA307O14.2
UniGene
Hs.486458
Gene Chromosome Location
Chr.6: 129576145-129710225 on Build GRCh38
Transcripts
NM_033515.2 XM_005267212.2 XM_005267213.1 XM_017011494.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3948004 c.1332C>G p.I444M Chr.6: 129605910-129605910 on Build GRCh38
6293984 c.1350C>A p.N450K Chr.6: 129605892-129605892 on Build GRCh38
4934212 c.1350C>T p.N450N Chr.6: 129605892-129605892 on Build GRCh38
7000306 c.1330A>G p.I444V Chr.6: 129605912-129605912 on Build GRCh38
7209529 c.1295A>T p.K432M Chr.6: 129605947-129605947 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539451121 Chr.6: 129605824-129605824 on Build GRCh38
rs981586849 Chr.6: 129605939-129605939 on Build GRCh38
rs765794990 Chr.6: 129605856-129605856 on Build GRCh38
rs917705561 Chr.6: 129605930-129605930 on Build GRCh38
rs147754877 Chr.6: 129605940-129605940 on Build GRCh38
rs781400499 Chr.6: 129605908-129605908 on Build GRCh38
rs775313640 Chr.6: 129605981-129605981 on Build GRCh38
rs202043590 Chr.6: 129605938-129605938 on Build GRCh38
rs145138074 Chr.6: 129605799-129605799 on Build GRCh38
rs972415026 Chr.6: 129605963-129605963 on Build GRCh38
rs370858739 Chr.6: 129605922-129605922 on Build GRCh38
rs1014698939 Chr.6: 129605929-129605929 on Build GRCh38
rs747202090 Chr.6: 129605970-129605972 on Build GRCh38
rs750958692 Chr.6: 129605868-129605868 on Build GRCh38
rs745383100 Chr.6: 129605962-129605962 on Build GRCh38
rs3813366 Chr.6: 129605869-129605869 on Build GRCh38
rs751720325 Chr.6: 129605888-129605888 on Build GRCh38
rs918247104 Chr.6: 129605978-129605978 on Build GRCh38
rs754197495 Chr.6: 129605832-129605832 on Build GRCh38
rs771783590 Chr.6: 129605973-129605973 on Build GRCh38
rs773967441 Chr.6: 129605943-129605943 on Build GRCh38
rs777653808 Chr.6: 129605857-129605857 on Build GRCh38
rs755138072 Chr.6: 129605898-129605898 on Build GRCh38
rs979647046 Chr.6: 129605802-129605802 on Build GRCh38
rs777550443 Chr.6: 129605923-129605923 on Build GRCh38
rs766518423 Chr.6: 129605877-129605877 on Build GRCh38
rs770345629 Chr.6: 129605942-129605942 on Build GRCh38
rs764538304 Chr.6: 129605831-129605831 on Build GRCh38

View Full Product Details