Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 68098523-68098782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG12-AS1
Entrez Gene ID
100289178
Gene Name
GNG12 antisense RNA 1
Gene Aliases
-
UniGene
Hs.677603
Gene Chromosome Location
Chr.1: 67832288-68202987 on Build GRCh38
Transcripts
NR_040077.1 NM_001002292.3 XM_011542191.2 XM_011542192.2

Gene Symbol
WLS
Entrez Gene ID
79971
Gene Name
wntless Wnt ligand secretion mediator
Gene Aliases
C1orf139, EVI, GPR177, MRP, mig-14
UniGene
Hs.647659
Gene Chromosome Location
Chr.1: 68098459-68233000 on Build GRCh38
Transcripts
NR_040077.1 NM_001002292.3 XM_011542191.2 XM_011542192.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
911856 c.1590G>A p.S530S Chr.1: 68098674-68098674 on Build GRCh38
2240443 c.1560G>A p.S520S Chr.1: 68098704-68098704 on Build GRCh38
911857 c.1559C>T p.S520L Chr.1: 68098705-68098705 on Build GRCh38
2240444 c.1555G>C p.V519L Chr.1: 68098709-68098709 on Build GRCh38
426635 c.1521C>T p.L507L Chr.1: 68098743-68098743 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776089916 Chr.1: 68098732-68098732 on Build GRCh38
rs758303393 Chr.1: 68098719-68098719 on Build GRCh38
rs765657108 Chr.1: 68098610-68098610 on Build GRCh38
rs138133925 Chr.1: 68098679-68098679 on Build GRCh38
rs777321300 Chr.1: 68098721-68098721 on Build GRCh38
rs760145246 Chr.1: 68098599-68098599 on Build GRCh38
rs766727080 Chr.1: 68098592-68098592 on Build GRCh38
rs770474601 Chr.1: 68098663-68098663 on Build GRCh38
rs112392383 Chr.1: 68098660-68098660 on Build GRCh38
rs3186845 Chr.1: 68098594-68098594 on Build GRCh38
rs748039646 Chr.1: 68098717-68098717 on Build GRCh38
rs767847835 Chr.1: 68098692-68098693 on Build GRCh38
rs903010968 Chr.1: 68098672-68098672 on Build GRCh38
rs751788591 Chr.1: 68098623-68098623 on Build GRCh38
rs373545096 Chr.1: 68098616-68098616 on Build GRCh38
rs372130459 Chr.1: 68098710-68098710 on Build GRCh38
rs550180906 Chr.1: 68098691-68098691 on Build GRCh38
rs1056166318 Chr.1: 68098606-68098606 on Build GRCh38
rs768709941 Chr.1: 68098638-68098638 on Build GRCh38
rs937750587 Chr.1: 68098588-68098588 on Build GRCh38
rs753403604 Chr.1: 68098706-68098709 on Build GRCh38
rs777118708 Chr.1: 68098674-68098674 on Build GRCh38
rs746716728 Chr.1: 68098656-68098656 on Build GRCh38
rs984998534 Chr.1: 68098564-68098564 on Build GRCh38
rs183122334 Chr.1: 68098709-68098709 on Build GRCh38
rs369009906 Chr.1: 68098555-68098555 on Build GRCh38
rs760155705 Chr.1: 68098691-68098691 on Build GRCh38
rs754129529 Chr.1: 68098598-68098598 on Build GRCh38
rs778759123 Chr.1: 68098645-68098645 on Build GRCh38
rs771522633 Chr.1: 68098649-68098649 on Build GRCh38
rs376791039 Chr.1: 68098624-68098624 on Build GRCh38
rs770620367 Chr.1: 68098729-68098729 on Build GRCh38
rs1023636544 Chr.1: 68098699-68098699 on Build GRCh38
rs764307335 Chr.1: 68098695-68098695 on Build GRCh38
rs200188571 Chr.1: 68098739-68098739 on Build GRCh38
rs940952640 Chr.1: 68098584-68098584 on Build GRCh38
rs747671564 Chr.1: 68098646-68098646 on Build GRCh38
rs776088338 Chr.1: 68098680-68098680 on Build GRCh38
rs187825885 Chr.1: 68098722-68098722 on Build GRCh38
rs751857880 Chr.1: 68098583-68098584 on Build GRCh38
rs141228692 Chr.1: 68098704-68098704 on Build GRCh38
rs144836484 Chr.1: 68098558-68098558 on Build GRCh38
rs116345986 Chr.1: 68098559-68098559 on Build GRCh38
rs530106043 Chr.1: 68098675-68098675 on Build GRCh38
rs749025614 Chr.1: 68098635-68098635 on Build GRCh38
rs561611966 Chr.1: 68098650-68098650 on Build GRCh38
rs779951950 Chr.1: 68098630-68098630 on Build GRCh38
rs200914637 Chr.1: 68098585-68098585 on Build GRCh38
rs1038532146 Chr.1: 68098562-68098562 on Build GRCh38
rs760971085 Chr.1: 68098591-68098591 on Build GRCh38
rs370144214 Chr.1: 68098705-68098705 on Build GRCh38
rs144056669 Chr.1: 68098702-68098702 on Build GRCh38
rs751571407 Chr.1: 68098696-68098696 on Build GRCh38
rs541758142 Chr.1: 68098617-68098617 on Build GRCh38
rs752828213 Chr.1: 68098614-68098614 on Build GRCh38

View Full Product Details