Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2270025-2270252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASCL2
Entrez Gene ID
430
Gene Name
achaete-scute family bHLH transcription factor 2
Gene Aliases
ASH2, HASH2, MASH2, bHLHa45
UniGene
Hs.152475
Gene Chromosome Location
Chr.11: 2268498-2270952 on Build GRCh38
Transcripts
NM_005170.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5703957 c.115C>T p.R39* Chr.11: 2270218-2270218 on Build GRCh38
6840516 c.207_213delCTTCCAG p.F70fs*15 Chr.11: 2270120-2270126 on Build GRCh38
6569968 c.273G>A p.T91T Chr.11: 2270060-2270060 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1002399430 Chr.11: 2270086-2270086 on Build GRCh38
rs759581492 Chr.11: 2270199-2270199 on Build GRCh38
rs1046893929 Chr.11: 2270065-2270065 on Build GRCh38
rs983743909 Chr.11: 2270217-2270217 on Build GRCh38
rs536039373 Chr.11: 2270235-2270235 on Build GRCh38
rs779339632 Chr.11: 2270144-2270144 on Build GRCh38
rs1054925019 Chr.11: 2270233-2270233 on Build GRCh38
rs941829629 Chr.11: 2270228-2270228 on Build GRCh38
rs755293969 Chr.11: 2270132-2270132 on Build GRCh38
rs778974931 Chr.11: 2270167-2270185 on Build GRCh38
rs776595190 Chr.11: 2270090-2270090 on Build GRCh38
rs976139581 Chr.11: 2270193-2270193 on Build GRCh38
rs762461460 Chr.11: 2270224-2270224 on Build GRCh38
rs772363556 Chr.11: 2270159-2270159 on Build GRCh38
rs767953362 Chr.11: 2270174-2270174 on Build GRCh38
rs780675695 Chr.11: 2270117-2270117 on Build GRCh38
rs1020628026 Chr.11: 2270104-2270104 on Build GRCh38
rs867992166 Chr.11: 2270154-2270154 on Build GRCh38
rs766337220 Chr.11: 2270076-2270076 on Build GRCh38
rs1046428802 Chr.11: 2270068-2270068 on Build GRCh38
rs964417466 Chr.11: 2270175-2270175 on Build GRCh38
rs372274549 Chr.11: 2270096-2270096 on Build GRCh38
rs756647017 Chr.11: 2270116-2270116 on Build GRCh38
rs771052351 Chr.11: 2270196-2270196 on Build GRCh38
rs763836932 Chr.11: 2270234-2270234 on Build GRCh38
rs940131231 Chr.11: 2270043-2270043 on Build GRCh38
rs764869919 Chr.11: 2270221-2270221 on Build GRCh38
rs908973960 Chr.11: 2270227-2270227 on Build GRCh38
rs906597790 Chr.11: 2270078-2270078 on Build GRCh38
rs902585147 Chr.11: 2270089-2270089 on Build GRCh38
rs920389089 Chr.11: 2270189-2270189 on Build GRCh38
rs983135224 Chr.11: 2270208-2270208 on Build GRCh38
rs868516654 Chr.11: 2270113-2270113 on Build GRCh38
rs1014003630 Chr.11: 2270053-2270053 on Build GRCh38
rs747062250 Chr.11: 2270184-2270184 on Build GRCh38
rs1007064104 Chr.11: 2270166-2270166 on Build GRCh38
rs758072033 Chr.11: 2270100-2270100 on Build GRCh38
rs748262851 Chr.11: 2270146-2270146 on Build GRCh38
rs773239491 Chr.11: 2270182-2270182 on Build GRCh38
rs775366015 Chr.11: 2270222-2270222 on Build GRCh38
rs950454006 Chr.11: 2270206-2270206 on Build GRCh38
rs866079637 Chr.11: 2270148-2270148 on Build GRCh38
rs749858000 Chr.11: 2270123-2270123 on Build GRCh38
rs374798855 Chr.11: 2270105-2270105 on Build GRCh38

View Full Product Details