Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63700441-63700698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFRP1
Entrez Gene ID
10139
Gene Name
ADP ribosylation factor related protein 1
Gene Aliases
ARL18, ARP, Arp1
UniGene
Hs.389277
Gene Chromosome Location
Chr.20: 63698642-63708013 on Build GRCh38
Transcripts
NM_001134758.3 NM_001267544.2 NM_001267545.2 NM_001267546.2 NM_001267547.2 NM_001267548.2 NM_001267549.2 NM_003224.5 NR_051954.2 NR_051955.2 NR_051956.2 NR_051957.2 NR_051958.2 XM_011528482.2 XM_011528483.1 XM_017027576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4662695 c.511delC p.L171fs*14 Chr.20: 63700609-63700609 on Build GRCh38
4662697 c.482G>A p.R161Q Chr.20: 63700638-63700638 on Build GRCh38
4789539 c.448G>T p.A150S Chr.20: 63700672-63700672 on Build GRCh38
6602963 c.369G>A p.P123P Chr.20: 63700462-63700462 on Build GRCh38
7139944 c.561C>A p.V187V Chr.20: 63700488-63700488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753227532 Chr.20: 63700483-63700483 on Build GRCh38
rs556108904 Chr.20: 63700556-63700556 on Build GRCh38
rs377188250 Chr.20: 63700554-63700554 on Build GRCh38
rs757399759 Chr.20: 63700610-63700610 on Build GRCh38
rs140213848 Chr.20: 63700462-63700462 on Build GRCh38
rs762341183 Chr.20: 63700642-63700642 on Build GRCh38
rs769854219 Chr.20: 63700491-63700491 on Build GRCh38
rs752492103 Chr.20: 63700463-63700465 on Build GRCh38
rs750696744 Chr.20: 63700674-63700674 on Build GRCh38
rs773068492 Chr.20: 63700639-63700639 on Build GRCh38
rs932562101 Chr.20: 63700477-63700477 on Build GRCh38
rs767914137 Chr.20: 63700465-63700465 on Build GRCh38
rs555052878 Chr.20: 63700594-63700594 on Build GRCh38
rs201441698 Chr.20: 63700509-63700509 on Build GRCh38
rs760205392 Chr.20: 63700468-63700468 on Build GRCh38
rs953852819 Chr.20: 63700619-63700619 on Build GRCh38
rs763805240 Chr.20: 63700471-63700471 on Build GRCh38
rs756142438 Chr.20: 63700646-63700646 on Build GRCh38
rs985172462 Chr.20: 63700576-63700576 on Build GRCh38
rs144341687 Chr.20: 63700523-63700523 on Build GRCh38
rs759473388 Chr.20: 63700648-63700648 on Build GRCh38
rs376358129 Chr.20: 63700656-63700656 on Build GRCh38
rs760520926 Chr.20: 63700665-63700665 on Build GRCh38
rs371879239 Chr.20: 63700564-63700564 on Build GRCh38
rs752515210 Chr.20: 63700599-63700599 on Build GRCh38
rs758569655 Chr.20: 63700467-63700467 on Build GRCh38
rs765959484 Chr.20: 63700645-63700645 on Build GRCh38
rs569900792 Chr.20: 63700565-63700565 on Build GRCh38
rs758302348 Chr.20: 63700523-63700524 on Build GRCh38
rs201543211 Chr.20: 63700613-63700613 on Build GRCh38
rs972592890 Chr.20: 63700663-63700663 on Build GRCh38
rs769076601 Chr.20: 63700625-63700625 on Build GRCh38
rs752431053 Chr.20: 63700664-63700664 on Build GRCh38
rs7267337 Chr.20: 63700581-63700581 on Build GRCh38
rs761386802 Chr.20: 63700521-63700521 on Build GRCh38
rs764223668 Chr.20: 63700479-63700479 on Build GRCh38
rs370122442 Chr.20: 63700555-63700555 on Build GRCh38
rs750436290 Chr.20: 63700487-63700487 on Build GRCh38
rs569223588 Chr.20: 63700464-63700464 on Build GRCh38
rs776120359 Chr.20: 63700561-63700561 on Build GRCh38
rs911763804 Chr.20: 63700572-63700572 on Build GRCh38
rs367735078 Chr.20: 63700520-63700520 on Build GRCh38
rs767291146 Chr.20: 63700598-63700598 on Build GRCh38
rs748541223 Chr.20: 63700634-63700634 on Build GRCh38
rs766357157 Chr.20: 63700543-63700543 on Build GRCh38
rs200743168 Chr.20: 63700653-63700653 on Build GRCh38
rs558842032 Chr.20: 63700567-63700567 on Build GRCh38
rs747503408 Chr.20: 63700621-63700621 on Build GRCh38
rs778118171 Chr.20: 63700493-63700493 on Build GRCh38
rs538542343 Chr.20: 63700566-63700566 on Build GRCh38
rs772401462 Chr.20: 63700560-63700560 on Build GRCh38
rs749986322 Chr.20: 63700530-63700530 on Build GRCh38
rs1003096421 Chr.20: 63700555-63700555 on Build GRCh38
rs149605591 Chr.20: 63700494-63700494 on Build GRCh38
rs774815023 Chr.20: 63700495-63700495 on Build GRCh38
rs759983214 Chr.20: 63700498-63700498 on Build GRCh38
rs373835704 Chr.20: 63700544-63700544 on Build GRCh38
rs35158527 Chr.20: 63700673-63700673 on Build GRCh38
rs778964903 Chr.20: 63700486-63700486 on Build GRCh38
rs1052533578 Chr.20: 63700672-63700672 on Build GRCh38
rs183956106 Chr.20: 63700676-63700676 on Build GRCh38
rs758568982 Chr.20: 63700617-63700617 on Build GRCh38
rs780392292 Chr.20: 63700620-63700620 on Build GRCh38
rs199647964 Chr.20: 63700580-63700580 on Build GRCh38
rs552866959 Chr.20: 63700490-63700490 on Build GRCh38
rs368664540 Chr.20: 63700542-63700542 on Build GRCh38
rs939300650 Chr.20: 63700655-63700655 on Build GRCh38
rs373533052 Chr.20: 63700514-63700514 on Build GRCh38
rs372715867 Chr.20: 63700591-63700591 on Build GRCh38
rs544032237 Chr.20: 63700463-63700463 on Build GRCh38
rs751124303 Chr.20: 63700590-63700591 on Build GRCh38
rs749558892 Chr.20: 63700546-63700546 on Build GRCh38
rs1021568713 Chr.20: 63700558-63700558 on Build GRCh38
rs1048665 Chr.20: 63700637-63700637 on Build GRCh38
rs532707971 Chr.20: 63700488-63700488 on Build GRCh38
rs948299995 Chr.20: 63700638-63700638 on Build GRCh38
rs367804866 Chr.20: 63700588-63700588 on Build GRCh38
rs775650276 Chr.20: 63700510-63700510 on Build GRCh38
rs200507208 Chr.20: 63700519-63700519 on Build GRCh38
rs961871779 Chr.20: 63700600-63700600 on Build GRCh38
rs535964457 Chr.20: 63700554-63700555 on Build GRCh38
rs563979293 Chr.20: 63700484-63700484 on Build GRCh38

View Full Product Details