Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 111791221-111791494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH2
Entrez Gene ID
217
Gene Name
aldehyde dehydrogenase 2 family (mitochondrial)
Gene Aliases
ALDH-E2, ALDHI, ALDM
UniGene
Hs.604551
Gene Chromosome Location
Chr.12: 111766887-111809985 on Build GRCh38
Transcripts
NM_000690.3 NM_001204889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3986639 c.686G>A p.G229D Chr.12: 111791310-111791310 on Build GRCh38
5504875 c.776C>T p.A259V Chr.12: 111791400-111791400 on Build GRCh38
4038783 c.741C>T p.A247A Chr.12: 111791365-111791365 on Build GRCh38
6598242 c.682G>A p.A228T Chr.12: 111791306-111791306 on Build GRCh38
1706284 c.752C>T p.S251F Chr.12: 111791376-111791376 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774511894 Chr.12: 111791470-111791470 on Build GRCh38
rs774642632 Chr.12: 111791350-111791350 on Build GRCh38
rs756375309 Chr.12: 111791281-111791281 on Build GRCh38
rs760546011 Chr.12: 111791366-111791366 on Build GRCh38
rs763423158 Chr.12: 111791338-111791338 on Build GRCh38
rs753791195 Chr.12: 111791379-111791379 on Build GRCh38
rs749694206 Chr.12: 111791336-111791336 on Build GRCh38
rs777767582 Chr.12: 111791280-111791280 on Build GRCh38
rs529768244 Chr.12: 111791333-111791333 on Build GRCh38
rs111277749 Chr.12: 111791365-111791365 on Build GRCh38
rs375522582 Chr.12: 111791440-111791440 on Build GRCh38
rs748550381 Chr.12: 111791327-111791327 on Build GRCh38
rs368395266 Chr.12: 111791450-111791450 on Build GRCh38
rs749950720 Chr.12: 111791391-111791391 on Build GRCh38
rs778333487 Chr.12: 111791285-111791285 on Build GRCh38
rs111540940 Chr.12: 111791373-111791373 on Build GRCh38
rs746303014 Chr.12: 111791438-111791438 on Build GRCh38
rs771771961 Chr.12: 111791317-111791317 on Build GRCh38
rs74595229 Chr.12: 111791277-111791277 on Build GRCh38
rs771107075 Chr.12: 111791343-111791343 on Build GRCh38
rs1001696628 Chr.12: 111791254-111791254 on Build GRCh38
rs756482078 Chr.12: 111791436-111791436 on Build GRCh38
rs1021095703 Chr.12: 111791297-111791297 on Build GRCh38
rs1034388083 Chr.12: 111791259-111791259 on Build GRCh38
rs771626523 Chr.12: 111791463-111791463 on Build GRCh38
rs781479853 Chr.12: 111791323-111791323 on Build GRCh38
rs868143172 Chr.12: 111791307-111791307 on Build GRCh38
rs893438603 Chr.12: 111791249-111791249 on Build GRCh38
rs543030829 Chr.12: 111791355-111791355 on Build GRCh38
rs544714733 Chr.12: 111791387-111791387 on Build GRCh38
rs925217597 Chr.12: 111791435-111791435 on Build GRCh38
rs909475464 Chr.12: 111791400-111791400 on Build GRCh38
rs757980778 Chr.12: 111791420-111791420 on Build GRCh38
rs759815398 Chr.12: 111791353-111791353 on Build GRCh38
rs538111819 Chr.12: 111791283-111791283 on Build GRCh38
rs778442341 Chr.12: 111791294-111791294 on Build GRCh38
rs752593972 Chr.12: 111791255-111791255 on Build GRCh38
rs745487886 Chr.12: 111791298-111791298 on Build GRCh38
rs139862632 Chr.12: 111791356-111791356 on Build GRCh38
rs748413982 Chr.12: 111791434-111791434 on Build GRCh38

View Full Product Details