Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50891705-50891976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADNP
Entrez Gene ID
23394
Gene Name
activity dependent neuroprotector homeobox
Gene Aliases
ADNP1, HVDAS, MRD28
UniGene
Hs.293736
Gene Chromosome Location
Chr.20: 50888918-50934984 on Build GRCh38
Transcripts
NM_001282531.1 NM_001282532.1 NM_015339.3 NM_181442.2 XM_011528747.2 XM_011528748.2 XM_017027757.1 XM_017027758.1 XM_017027759.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7170642 c.2917G>C p.E973Q Chr.20: 50891797-50891797 on Build GRCh38
5916807 c.2897A>T p.K966I Chr.20: 50891817-50891817 on Build GRCh38
6539453 c.2919G>C p.E973D Chr.20: 50891795-50891795 on Build GRCh38
1681758 c.2747T>G p.I916S Chr.20: 50891967-50891967 on Build GRCh38
7078980 c.2979A>G p.P993P Chr.20: 50891735-50891735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780569502 Chr.20: 50891747-50891747 on Build GRCh38
rs564373258 Chr.20: 50891743-50891743 on Build GRCh38
rs746768340 Chr.20: 50891886-50891886 on Build GRCh38
rs768463704 Chr.20: 50891889-50891889 on Build GRCh38
rs748421305 Chr.20: 50891859-50891859 on Build GRCh38
rs750133102 Chr.20: 50891824-50891826 on Build GRCh38
rs143103063 Chr.20: 50891771-50891771 on Build GRCh38
rs749431988 Chr.20: 50891870-50891870 on Build GRCh38
rs767933419 Chr.20: 50891930-50891930 on Build GRCh38
rs745827301 Chr.20: 50891772-50891772 on Build GRCh38
rs141643081 Chr.20: 50891792-50891792 on Build GRCh38
rs6096164 Chr.20: 50891812-50891812 on Build GRCh38
rs764863978 Chr.20: 50891900-50891900 on Build GRCh38
rs184697035 Chr.20: 50891905-50891905 on Build GRCh38
rs777909404 Chr.20: 50891864-50891864 on Build GRCh38
rs772068264 Chr.20: 50891773-50891773 on Build GRCh38
rs371901809 Chr.20: 50891946-50891946 on Build GRCh38
rs76458950 Chr.20: 50891932-50891932 on Build GRCh38
rs764817280 Chr.20: 50891822-50891822 on Build GRCh38
rs143425387 Chr.20: 50891929-50891929 on Build GRCh38
rs6096165 Chr.20: 50891846-50891846 on Build GRCh38
rs752139612 Chr.20: 50891926-50891926 on Build GRCh38
rs771978437 Chr.20: 50891873-50891873 on Build GRCh38
rs201922657 Chr.20: 50891833-50891833 on Build GRCh38
rs755785720 Chr.20: 50891830-50891832 on Build GRCh38
rs776090032 Chr.20: 50891895-50891895 on Build GRCh38
rs367605121 Chr.20: 50891858-50891858 on Build GRCh38
rs531230573 Chr.20: 50891746-50891746 on Build GRCh38
rs144684998 Chr.20: 50891783-50891783 on Build GRCh38
rs946407237 Chr.20: 50891920-50891920 on Build GRCh38
rs913242351 Chr.20: 50891829-50891829 on Build GRCh38
rs750828266 Chr.20: 50891831-50891831 on Build GRCh38
rs587777525 Chr.20: 50891906-50891906 on Build GRCh38
rs373951274 Chr.20: 50891904-50891904 on Build GRCh38
rs779248718 Chr.20: 50891745-50891745 on Build GRCh38
rs769945500 Chr.20: 50891770-50891770 on Build GRCh38
rs766959422 Chr.20: 50891906-50891906 on Build GRCh38
rs1036107284 Chr.20: 50891951-50891951 on Build GRCh38
rs370874843 Chr.20: 50891883-50891883 on Build GRCh38
rs747443821 Chr.20: 50891748-50891748 on Build GRCh38
rs766657056 Chr.20: 50891841-50891846 on Build GRCh38
rs751947530 Chr.20: 50891849-50891849 on Build GRCh38
rs568101760 Chr.20: 50891754-50891754 on Build GRCh38
rs994682969 Chr.20: 50891785-50891785 on Build GRCh38
rs748142440 Chr.20: 50891763-50891763 on Build GRCh38
rs551485241 Chr.20: 50891878-50891878 on Build GRCh38
rs534353954 Chr.20: 50891834-50891834 on Build GRCh38
rs756086209 Chr.20: 50891863-50891863 on Build GRCh38
rs763707378 Chr.20: 50891793-50891793 on Build GRCh38
rs148496595 Chr.20: 50891942-50891942 on Build GRCh38
rs201104498 Chr.20: 50891899-50891899 on Build GRCh38
rs756349173 Chr.20: 50891934-50891934 on Build GRCh38
rs187266199 Chr.20: 50891751-50891751 on Build GRCh38
rs761180077 Chr.20: 50891813-50891813 on Build GRCh38
rs767049382 Chr.20: 50891837-50891837 on Build GRCh38
rs1014546816 Chr.20: 50891729-50891729 on Build GRCh38

View Full Product Details