Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 123163769-123164034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUB3
Entrez Gene ID
9184
Gene Name
BUB3, mitotic checkpoint protein
Gene Aliases
BUB3L, hBUB3
UniGene
Hs.418533
Gene Chromosome Location
Chr.10: 123154244-123165370 on Build GRCh38
Transcripts
NM_001007793.2 NM_004725.3

SNP Detail

SNP ID Chromosome Location
rs902226028 Chr.10: 123163947-123163947 on Build GRCh38
rs374733234 Chr.10: 123163883-123163883 on Build GRCh38
rs975315228 Chr.10: 123163984-123163987 on Build GRCh38
rs1053658348 Chr.10: 123163983-123163983 on Build GRCh38
rs995039047 Chr.10: 123163889-123163889 on Build GRCh38
rs767568564 Chr.10: 123163818-123163818 on Build GRCh38
rs921206630 Chr.10: 123163994-123163997 on Build GRCh38
rs200759442 Chr.10: 123163983-123163984 on Build GRCh38
rs771143104 Chr.10: 123163943-123163945 on Build GRCh38
rs546143319 Chr.10: 123163790-123163790 on Build GRCh38
rs1012201007 Chr.10: 123164004-123164004 on Build GRCh38
rs547618270 Chr.10: 123163969-123163969 on Build GRCh38
rs529436507 Chr.10: 123163922-123163922 on Build GRCh38
rs747621370 Chr.10: 123163884-123163884 on Build GRCh38
rs75406601 Chr.10: 123163993-123163994 on Build GRCh38
rs566010640 Chr.10: 123163995-123163995 on Build GRCh38
rs750383253 Chr.10: 123163822-123163822 on Build GRCh38
rs562085562 Chr.10: 123163921-123163921 on Build GRCh38
rs775015692 Chr.10: 123163827-123163827 on Build GRCh38
rs201226325 Chr.10: 123163877-123163877 on Build GRCh38
rs978653301 Chr.10: 123163999-123164002 on Build GRCh38
rs116437658 Chr.10: 123163813-123163813 on Build GRCh38
rs756028806 Chr.10: 123163861-123163861 on Build GRCh38
rs1042139768 Chr.10: 123163929-123163929 on Build GRCh38
rs1000612064 Chr.10: 123163967-123163967 on Build GRCh38
rs749009869 Chr.10: 123163876-123163876 on Build GRCh38
rs965055647 Chr.10: 123163984-123163984 on Build GRCh38
rs543848746 Chr.10: 123163911-123163911 on Build GRCh38
rs777005250 Chr.10: 123163933-123163933 on Build GRCh38
rs781723991 Chr.10: 123163949-123163951 on Build GRCh38
rs951195366 Chr.10: 123163914-123163917 on Build GRCh38
rs201333657 Chr.10: 123163871-123163871 on Build GRCh38

View Full Product Details