Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 4982702-4982967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BHLHE40
Entrez Gene ID
8553
Gene Name
basic helix-loop-helix family member e40
Gene Aliases
BHLHB2, DEC1, HLHB2, SHARP-2, STRA13, Stra14
UniGene
Hs.744856
Gene Chromosome Location
Chr.3: 4979412-4985181 on Build GRCh38
Transcripts
NM_003670.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1046428 c.506C>T p.S169L Chr.3: 4982959-4982959 on Build GRCh38
6848440 c.466_490del25 p.Y156fs*3 Chr.3: 4982919-4982943 on Build GRCh38
6611458 c.398G>A p.R133K Chr.3: 4982851-4982851 on Build GRCh38
4118717 c.490C>G p.R164G Chr.3: 4982943-4982943 on Build GRCh38
4665677 c.452G>A p.R151Q Chr.3: 4982905-4982905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs56138530 Chr.3: 4982741-4982742 on Build GRCh38
rs776019216 Chr.3: 4982879-4982879 on Build GRCh38
rs11398309 Chr.3: 4982730-4982731 on Build GRCh38
rs544708561 Chr.3: 4982913-4982913 on Build GRCh38
rs1007462656 Chr.3: 4982821-4982821 on Build GRCh38
rs760004090 Chr.3: 4982846-4982846 on Build GRCh38
rs200395269 Chr.3: 4982876-4982876 on Build GRCh38
rs771192581 Chr.3: 4982830-4982830 on Build GRCh38
rs371023207 Chr.3: 4982817-4982817 on Build GRCh38
rs779263807 Chr.3: 4982934-4982934 on Build GRCh38
rs1019908752 Chr.3: 4982770-4982770 on Build GRCh38
rs754866445 Chr.3: 4982910-4982910 on Build GRCh38
rs367916203 Chr.3: 4982865-4982865 on Build GRCh38
rs537778709 Chr.3: 4982802-4982802 on Build GRCh38
rs746954581 Chr.3: 4982788-4982788 on Build GRCh38
rs756156275 Chr.3: 4982926-4982926 on Build GRCh38
rs539171296 Chr.3: 4982806-4982806 on Build GRCh38
rs754810290 Chr.3: 4982807-4982807 on Build GRCh38
rs551273788 Chr.3: 4982731-4982732 on Build GRCh38
rs933649148 Chr.3: 4982772-4982772 on Build GRCh38
rs764676216 Chr.3: 4982894-4982894 on Build GRCh38
rs774521888 Chr.3: 4982831-4982831 on Build GRCh38
rs533078309 Chr.3: 4982731-4982731 on Build GRCh38
rs148401238 Chr.3: 4982896-4982896 on Build GRCh38
rs201532039 Chr.3: 4982808-4982808 on Build GRCh38
rs770050588 Chr.3: 4982812-4982812 on Build GRCh38
rs371986656 Chr.3: 4982904-4982904 on Build GRCh38
rs999979586 Chr.3: 4982784-4982784 on Build GRCh38
rs772560334 Chr.3: 4982795-4982795 on Build GRCh38
rs199844835 Chr.3: 4982742-4982742 on Build GRCh38
rs146720298 Chr.3: 4982944-4982944 on Build GRCh38
rs139606133 Chr.3: 4982897-4982897 on Build GRCh38
rs752635944 Chr.3: 4982920-4982920 on Build GRCh38
rs758068591 Chr.3: 4982796-4982796 on Build GRCh38
rs779672732 Chr.3: 4982797-4982797 on Build GRCh38
rs904169835 Chr.3: 4982780-4982780 on Build GRCh38
rs200306552 Chr.3: 4982941-4982941 on Build GRCh38
rs972664279 Chr.3: 4982741-4982741 on Build GRCh38
rs375227436 Chr.3: 4982918-4982918 on Build GRCh38
rs552802969 Chr.3: 4982909-4982909 on Build GRCh38
rs779995075 Chr.3: 4982752-4982752 on Build GRCh38
rs368425219 Chr.3: 4982933-4982933 on Build GRCh38
rs750123092 Chr.3: 4982795-4982795 on Build GRCh38
rs140413407 Chr.3: 4982819-4982819 on Build GRCh38
rs777935965 Chr.3: 4982931-4982931 on Build GRCh38
rs761289332 Chr.3: 4982891-4982891 on Build GRCh38
rs934181679 Chr.3: 4982756-4982756 on Build GRCh38

View Full Product Details