Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48956133-48956406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAX
Entrez Gene ID
581
Gene Name
BCL2 associated X, apoptosis regulator
Gene Aliases
BCL2L4
UniGene
Hs.624291
Gene Chromosome Location
Chr.19: 48954825-48961798 on Build GRCh38
Transcripts
NM_001291428.1 NM_001291429.1 NM_001291430.1 NM_001291431.1 NM_004324.3 NM_138761.3 NM_138763.3 NM_138764.4 NR_027882.1 XM_017027077.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1590199 c.266G>A p.R89Q Chr.19: 48956230-48956230 on Build GRCh38
6277247 c.234-4G>T p.? Chr.19: 48956194-48956194 on Build GRCh38
3108292 c.247G>A p.V83M Chr.19: 48956211-48956211 on Build GRCh38
6085168 c.353G>T p.S118I Chr.19: 48956317-48956317 on Build GRCh38
3108296 c.260delC p.R89fs*44 Chr.19: 48956224-48956224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781054379 Chr.19: 48956185-48956185 on Build GRCh38
rs774010704 Chr.19: 48956386-48956386 on Build GRCh38
rs765985987 Chr.19: 48956317-48956317 on Build GRCh38
rs772339112 Chr.19: 48956258-48956258 on Build GRCh38
rs374052135 Chr.19: 48956228-48956228 on Build GRCh38
rs759655007 Chr.19: 48956376-48956376 on Build GRCh38
rs899560616 Chr.19: 48956277-48956277 on Build GRCh38
rs201578495 Chr.19: 48956297-48956297 on Build GRCh38
rs367558446 Chr.19: 48956283-48956283 on Build GRCh38
rs756035440 Chr.19: 48956365-48956365 on Build GRCh38
rs749142585 Chr.19: 48956245-48956245 on Build GRCh38
rs777326329 Chr.19: 48956211-48956211 on Build GRCh38
rs867504612 Chr.19: 48956314-48956314 on Build GRCh38
rs765725612 Chr.19: 48956224-48956224 on Build GRCh38
rs754460156 Chr.19: 48956341-48956341 on Build GRCh38
rs760733803 Chr.19: 48956208-48956208 on Build GRCh38
rs777236472 Chr.19: 48956282-48956282 on Build GRCh38
rs747228737 Chr.19: 48956377-48956377 on Build GRCh38
rs148204524 Chr.19: 48956270-48956270 on Build GRCh38
rs757702927 Chr.19: 48956154-48956154 on Build GRCh38
rs780275950 Chr.19: 48956373-48956373 on Build GRCh38
rs967900060 Chr.19: 48956235-48956235 on Build GRCh38
rs974671518 Chr.19: 48956348-48956348 on Build GRCh38
rs371147985 Chr.19: 48956193-48956193 on Build GRCh38
rs749045019 Chr.19: 48956367-48956367 on Build GRCh38
rs777758516 Chr.19: 48956366-48956366 on Build GRCh38
rs778652291 Chr.19: 48956155-48956155 on Build GRCh38
rs754446832 Chr.19: 48956229-48956229 on Build GRCh38
rs370634054 Chr.19: 48956210-48956210 on Build GRCh38
rs202190487 Chr.19: 48956230-48956230 on Build GRCh38
rs768620338 Chr.19: 48956175-48956175 on Build GRCh38
rs769766616 Chr.19: 48956237-48956237 on Build GRCh38
rs775579234 Chr.19: 48956261-48956261 on Build GRCh38
rs762222733 Chr.19: 48956212-48956212 on Build GRCh38
rs869084780 Chr.19: 48956302-48956303 on Build GRCh38
rs765892616 Chr.19: 48956292-48956292 on Build GRCh38
rs759725888 Chr.19: 48956200-48956200 on Build GRCh38
rs751182192 Chr.19: 48956339-48956339 on Build GRCh38
rs144896180 Chr.19: 48956353-48956353 on Build GRCh38
rs1010232888 Chr.19: 48956368-48956369 on Build GRCh38
rs750121337 Chr.19: 48956160-48956160 on Build GRCh38
rs376411979 Chr.19: 48956172-48956172 on Build GRCh38
rs779583892 Chr.19: 48956168-48956168 on Build GRCh38
rs769587531 Chr.19: 48956190-48956190 on Build GRCh38
rs777525298 Chr.19: 48956241-48956241 on Build GRCh38
rs776628729 Chr.19: 48956279-48956292 on Build GRCh38
rs775629755 Chr.19: 48956207-48956207 on Build GRCh38
rs752257224 Chr.19: 48956357-48956357 on Build GRCh38
rs763289526 Chr.19: 48956301-48956301 on Build GRCh38
rs747867623 Chr.19: 48956189-48956189 on Build GRCh38
rs1051627684 Chr.19: 48956375-48956375 on Build GRCh38
rs371363325 Chr.19: 48956372-48956372 on Build GRCh38
rs979919138 Chr.19: 48956162-48956162 on Build GRCh38
rs867104644 Chr.19: 48956294-48956294 on Build GRCh38
rs964044483 Chr.19: 48956346-48956346 on Build GRCh38
rs768643044 Chr.19: 48956271-48956271 on Build GRCh38
rs772234712 Chr.19: 48956201-48956201 on Build GRCh38
rs757940370 Chr.19: 48956165-48956165 on Build GRCh38

View Full Product Details