Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 26915843-26916082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASUN
Entrez Gene ID
55726
Gene Name
asunder, spermatogenesis regulator
Gene Aliases
C12orf11, GCT1, Mat89Bb, NET48, SPATA30
UniGene
Hs.505077
Gene Chromosome Location
Chr.12: 26905171-26938326 on Build GRCh38
Transcripts
NM_018164.2 XM_005253422.3 XM_011520750.2 XM_011520751.2 XM_011520752.2 XM_011520753.2 XM_017019632.1 XM_017019633.1 XM_017019634.1 XM_017019635.1 XM_017019636.1 XM_017019637.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
226713 c.1181C>T p.S394F Chr.12: 26916069-26916069 on Build GRCh38
3792471 c.1234G>A p.D412N Chr.12: 26916016-26916016 on Build GRCh38
6606250 c.1178G>A p.R393Q Chr.12: 26916072-26916072 on Build GRCh38
430947 c.1242G>C p.R414R Chr.12: 26916008-26916008 on Build GRCh38
1361050 c.1177C>T p.R393* Chr.12: 26916073-26916073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893183452 Chr.12: 26916051-26916051 on Build GRCh38
rs769601793 Chr.12: 26916003-26916003 on Build GRCh38
rs981933955 Chr.12: 26915945-26915945 on Build GRCh38
rs568200794 Chr.12: 26916041-26916041 on Build GRCh38
rs779131364 Chr.12: 26915957-26915957 on Build GRCh38
rs757812846 Chr.12: 26915877-26915877 on Build GRCh38
rs923795657 Chr.12: 26915967-26915967 on Build GRCh38
rs965338689 Chr.12: 26915944-26915944 on Build GRCh38
rs761769634 Chr.12: 26915876-26915876 on Build GRCh38
rs772130012 Chr.12: 26915962-26915962 on Build GRCh38
rs35037404 Chr.12: 26916050-26916050 on Build GRCh38
rs949291871 Chr.12: 26915938-26915938 on Build GRCh38
rs1017824941 Chr.12: 26915939-26915939 on Build GRCh38
rs916530767 Chr.12: 26915910-26915910 on Build GRCh38
rs374930017 Chr.12: 26916002-26916002 on Build GRCh38
rs764081535 Chr.12: 26916056-26916056 on Build GRCh38
rs759214384 Chr.12: 26915980-26915980 on Build GRCh38
rs952484011 Chr.12: 26915909-26915909 on Build GRCh38
rs762850300 Chr.12: 26916053-26916053 on Build GRCh38
rs976070267 Chr.12: 26915981-26915981 on Build GRCh38
rs750866673 Chr.12: 26915963-26915963 on Build GRCh38
rs1026700429 Chr.12: 26915877-26915879 on Build GRCh38
rs781380615 Chr.12: 26915960-26915960 on Build GRCh38

View Full Product Details