Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 35717773-35718042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOL5
Entrez Gene ID
80831
Gene Name
apolipoprotein L5
Gene Aliases
APOL-V, APOLV
UniGene
Hs.326561
Gene Chromosome Location
Chr.22: 35708424-35730279 on Build GRCh38
Transcripts
NM_030642.1 XM_006724321.3 XM_017028945.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5455018 c.4C>T p.P2S Chr.22: 35717875-35717875 on Build GRCh38
6602457 c.55+1G>A p.? Chr.22: 35717927-35717927 on Build GRCh38
5909861 c.5C>T p.P2L Chr.22: 35717876-35717876 on Build GRCh38
3553890 c.11G>A p.G4D Chr.22: 35717882-35717882 on Build GRCh38
299060 c.22A>C p.N8H Chr.22: 35717893-35717893 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780271766 Chr.22: 35717858-35717858 on Build GRCh38
rs959117339 Chr.22: 35717836-35717836 on Build GRCh38
rs758498118 Chr.22: 35717966-35717966 on Build GRCh38
rs763815526 Chr.22: 35717880-35717880 on Build GRCh38
rs752055355 Chr.22: 35717825-35717825 on Build GRCh38
rs756482337 Chr.22: 35717857-35717857 on Build GRCh38
rs764712902 Chr.22: 35717985-35717985 on Build GRCh38
rs987318809 Chr.22: 35718019-35718019 on Build GRCh38
rs750764405 Chr.22: 35717855-35717855 on Build GRCh38
rs765039172 Chr.22: 35717899-35717899 on Build GRCh38
rs766669140 Chr.22: 35717932-35717932 on Build GRCh38
rs757825632 Chr.22: 35717831-35717831 on Build GRCh38
rs778954997 Chr.22: 35717870-35717870 on Build GRCh38
rs779147734 Chr.22: 35717952-35717952 on Build GRCh38
rs568782885 Chr.22: 35717957-35717957 on Build GRCh38
rs771966312 Chr.22: 35717877-35717877 on Build GRCh38
rs202132559 Chr.22: 35717967-35717967 on Build GRCh38
rs750913785 Chr.22: 35717916-35717916 on Build GRCh38
rs201209006 Chr.22: 35717907-35717907 on Build GRCh38
rs142982706 Chr.22: 35717948-35717948 on Build GRCh38
rs991653234 Chr.22: 35717933-35717933 on Build GRCh38
rs188546228 Chr.22: 35717881-35717881 on Build GRCh38
rs773074929 Chr.22: 35717878-35717878 on Build GRCh38
rs2899256 Chr.22: 35718007-35718007 on Build GRCh38
rs368647027 Chr.22: 35717910-35717910 on Build GRCh38
rs762529215 Chr.22: 35717854-35717854 on Build GRCh38
rs754996683 Chr.22: 35717864-35717864 on Build GRCh38
rs759274718 Chr.22: 35717898-35717898 on Build GRCh38
rs536770262 Chr.22: 35717888-35717888 on Build GRCh38
rs4821438 Chr.22: 35718000-35718000 on Build GRCh38
rs971794461 Chr.22: 35717991-35717991 on Build GRCh38
rs899817073 Chr.22: 35718009-35718009 on Build GRCh38
rs376655085 Chr.22: 35717947-35717947 on Build GRCh38
rs945170559 Chr.22: 35717917-35717917 on Build GRCh38
rs371739052 Chr.22: 35717919-35717919 on Build GRCh38
rs776279359 Chr.22: 35717882-35717882 on Build GRCh38
rs762588197 Chr.22: 35717908-35717908 on Build GRCh38
rs974145681 Chr.22: 35717809-35717809 on Build GRCh38
rs757947101 Chr.22: 35717884-35717884 on Build GRCh38
rs903188434 Chr.22: 35717804-35717804 on Build GRCh38
rs768202526 Chr.22: 35717834-35717834 on Build GRCh38
rs746924095 Chr.22: 35717971-35717971 on Build GRCh38
rs748295040 Chr.22: 35717876-35717876 on Build GRCh38

View Full Product Details