Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29729473-29729698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABP7
Entrez Gene ID
164633
Gene Name
calcium binding protein 7
Gene Aliases
CALN2
UniGene
Hs.643608
Gene Chromosome Location
Chr.22: 29720090-29731833 on Build GRCh38
Transcripts
NM_182527.2 XM_006724161.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7298937 c.580G>A p.A194T Chr.22: 29729501-29729501 on Build GRCh38
4642615 c.592A>G p.I198V Chr.22: 29729513-29729513 on Build GRCh38
7188528 c.600T>G p.S200R Chr.22: 29729521-29729521 on Build GRCh38
5859144 c.614C>T p.A205V Chr.22: 29729535-29729535 on Build GRCh38
1682191 c.591C>A p.F197L Chr.22: 29729512-29729512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs5997507 Chr.22: 29729607-29729607 on Build GRCh38
rs894069201 Chr.22: 29729507-29729507 on Build GRCh38
rs771598397 Chr.22: 29729613-29729613 on Build GRCh38
rs563687450 Chr.22: 29729653-29729653 on Build GRCh38
rs374153706 Chr.22: 29729608-29729608 on Build GRCh38
rs757392012 Chr.22: 29729594-29729594 on Build GRCh38
rs552053465 Chr.22: 29729668-29729668 on Build GRCh38
rs541654626 Chr.22: 29729619-29729619 on Build GRCh38
rs373895667 Chr.22: 29729536-29729536 on Build GRCh38
rs570456067 Chr.22: 29729675-29729675 on Build GRCh38
rs777605216 Chr.22: 29729563-29729565 on Build GRCh38
rs748981553 Chr.22: 29729523-29729523 on Build GRCh38
rs377507972 Chr.22: 29729595-29729595 on Build GRCh38
rs530840725 Chr.22: 29729654-29729654 on Build GRCh38
rs773977546 Chr.22: 29729532-29729532 on Build GRCh38
rs200183587 Chr.22: 29729501-29729501 on Build GRCh38
rs780525427 Chr.22: 29729601-29729601 on Build GRCh38
rs751729168 Chr.22: 29729586-29729586 on Build GRCh38
rs780216066 Chr.22: 29729513-29729513 on Build GRCh38
rs764218505 Chr.22: 29729574-29729574 on Build GRCh38
rs977764394 Chr.22: 29729656-29729656 on Build GRCh38
rs936395689 Chr.22: 29729667-29729667 on Build GRCh38
rs879145418 Chr.22: 29729676-29729676 on Build GRCh38
rs868543347 Chr.22: 29729559-29729559 on Build GRCh38
rs756519035 Chr.22: 29729600-29729600 on Build GRCh38
rs773151916 Chr.22: 29729546-29729546 on Build GRCh38
rs377460912 Chr.22: 29729494-29729494 on Build GRCh38
rs201111876 Chr.22: 29729495-29729495 on Build GRCh38
rs959655868 Chr.22: 29729630-29729630 on Build GRCh38
rs780840672 Chr.22: 29729577-29729578 on Build GRCh38
rs746286699 Chr.22: 29729506-29729506 on Build GRCh38
rs925049420 Chr.22: 29729666-29729666 on Build GRCh38
rs772042088 Chr.22: 29729542-29729542 on Build GRCh38
rs146484285 Chr.22: 29729571-29729571 on Build GRCh38
rs766203600 Chr.22: 29729552-29729552 on Build GRCh38
rs5997508 Chr.22: 29729631-29729631 on Build GRCh38
rs200220807 Chr.22: 29729596-29729596 on Build GRCh38
rs1008557029 Chr.22: 29729555-29729555 on Build GRCh38
rs763297168 Chr.22: 29729573-29729573 on Build GRCh38
rs377500141 Chr.22: 29729614-29729614 on Build GRCh38
rs747747030 Chr.22: 29729610-29729610 on Build GRCh38
rs760622867 Chr.22: 29729615-29729615 on Build GRCh38
rs746663947 Chr.22: 29729576-29729577 on Build GRCh38
rs768181054 Chr.22: 29729530-29729530 on Build GRCh38
rs565605720 Chr.22: 29729562-29729562 on Build GRCh38
rs760568088 Chr.22: 29729548-29729548 on Build GRCh38
rs770306080 Chr.22: 29729509-29729509 on Build GRCh38
rs559839645 Chr.22: 29729627-29729627 on Build GRCh38
rs371482198 Chr.22: 29729500-29729500 on Build GRCh38
rs1009794420 Chr.22: 29729520-29729520 on Build GRCh38
rs530234716 Chr.22: 29729628-29729628 on Build GRCh38

View Full Product Details