Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 75564207-75564444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf26
Entrez Gene ID
152816
Gene Name
chromosome 4 open reading frame 26
Gene Aliases
AI2A4
UniGene
Hs.24510
Gene Chromosome Location
Chr.4: 75554997-75565893 on Build GRCh38
Transcripts
NM_001206981.1 NM_001257072.1 NM_178497.3 NR_046429.1 NR_046430.1 XM_011531668.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3605846 c.288C>T p.S96S Chr.4: 75564290-75564290 on Build GRCh38
3918139 c.274C>T p.P92S Chr.4: 75564320-75564320 on Build GRCh38
2831655 c.409A>G p.R137G Chr.4: 75564411-75564411 on Build GRCh38
5629738 c.291T>G p.I97M Chr.4: 75564293-75564293 on Build GRCh38
5527768 c.342C>T p.F114F Chr.4: 75564344-75564344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150692328 Chr.4: 75564253-75564253 on Build GRCh38
rs772937947 Chr.4: 75564363-75564363 on Build GRCh38
rs1038145906 Chr.4: 75564368-75564368 on Build GRCh38
rs2306174 Chr.4: 75564372-75564372 on Build GRCh38
rs144879318 Chr.4: 75564254-75564254 on Build GRCh38
rs1000519307 Chr.4: 75564375-75564375 on Build GRCh38
rs371428561 Chr.4: 75564267-75564267 on Build GRCh38
rs946796697 Chr.4: 75564311-75564311 on Build GRCh38
rs765343106 Chr.4: 75564274-75564274 on Build GRCh38
rs746286264 Chr.4: 75564406-75564406 on Build GRCh38
rs76243433 Chr.4: 75564265-75564265 on Build GRCh38
rs762972493 Chr.4: 75564251-75564251 on Build GRCh38
rs748978797 Chr.4: 75564420-75564420 on Build GRCh38
rs767321689 Chr.4: 75564361-75564361 on Build GRCh38
rs758475707 Chr.4: 75564282-75564282 on Build GRCh38
rs778192571 Chr.4: 75564286-75564286 on Build GRCh38
rs540030139 Chr.4: 75564318-75564318 on Build GRCh38
rs543104609 Chr.4: 75564382-75564382 on Build GRCh38
rs765386366 Chr.4: 75564398-75564400 on Build GRCh38
rs765305769 Chr.4: 75564371-75564371 on Build GRCh38
rs150408808 Chr.4: 75564407-75564407 on Build GRCh38
rs745542848 Chr.4: 75564233-75564233 on Build GRCh38
rs563697262 Chr.4: 75564255-75564255 on Build GRCh38
rs867328692 Chr.4: 75564350-75564350 on Build GRCh38
rs149011730 Chr.4: 75564276-75564276 on Build GRCh38
rs1044490985 Chr.4: 75564284-75564284 on Build GRCh38
rs202035661 Chr.4: 75564232-75564232 on Build GRCh38
rs774389402 Chr.4: 75564348-75564348 on Build GRCh38
rs142989792 Chr.4: 75564230-75564230 on Build GRCh38
rs138123570 Chr.4: 75564423-75564423 on Build GRCh38
rs752032704 Chr.4: 75564385-75564385 on Build GRCh38
rs769319224 Chr.4: 75564238-75564238 on Build GRCh38
rs761879769 Chr.4: 75564351-75564351 on Build GRCh38
rs377354901 Chr.4: 75564421-75564421 on Build GRCh38
rs775135597 Chr.4: 75564326-75564326 on Build GRCh38
rs531246901 Chr.4: 75564317-75564317 on Build GRCh38
rs369987443 Chr.4: 75564354-75564354 on Build GRCh38
rs774915989 Chr.4: 75564250-75564250 on Build GRCh38
rs142537029 Chr.4: 75564340-75564340 on Build GRCh38
rs781447816 Chr.4: 75564400-75564400 on Build GRCh38
rs748742029 Chr.4: 75564339-75564339 on Build GRCh38
rs866941536 Chr.4: 75564275-75564275 on Build GRCh38
rs146645381 Chr.4: 75564364-75564364 on Build GRCh38
rs140756390 Chr.4: 75564323-75564323 on Build GRCh38
rs373107558 Chr.4: 75564412-75564412 on Build GRCh38
rs761607700 Chr.4: 75564241-75564241 on Build GRCh38
rs767246401 Chr.4: 75564263-75564263 on Build GRCh38
rs115202810 Chr.4: 75564374-75564374 on Build GRCh38
rs781610477 Chr.4: 75564402-75564402 on Build GRCh38
rs747540711 Chr.4: 75564316-75564316 on Build GRCh38
rs764826666 Chr.4: 75564264-75564264 on Build GRCh38
rs184413027 Chr.4: 75564321-75564321 on Build GRCh38

View Full Product Details