Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58113892-58114153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP20
Entrez Gene ID
29105
Gene Name
cilia and flagella associated protein 20
Gene Aliases
C16orf80, EVORF, GTL3, fSAP23
UniGene
Hs.532755
Gene Chromosome Location
Chr.16: 58113588-58129425 on Build GRCh38
Transcripts
NM_013242.2 NR_136632.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
289566 c.579A>G p.Q193Q Chr.16: 58114028-58114028 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1034449787 Chr.16: 58113963-58113963 on Build GRCh38
rs929532034 Chr.16: 58114115-58114115 on Build GRCh38
rs985594789 Chr.16: 58114055-58114055 on Build GRCh38
rs981843775 Chr.16: 58114125-58114125 on Build GRCh38
rs188242443 Chr.16: 58114108-58114108 on Build GRCh38
rs764195180 Chr.16: 58114056-58114056 on Build GRCh38
rs761282569 Chr.16: 58114061-58114061 on Build GRCh38
rs910553745 Chr.16: 58114071-58114071 on Build GRCh38
rs71850787 Chr.16: 58113996-58113996 on Build GRCh38
rs201886107 Chr.16: 58114010-58114010 on Build GRCh38
rs375016474 Chr.16: 58113955-58113955 on Build GRCh38
rs566869745 Chr.16: 58114086-58114086 on Build GRCh38
rs774617353 Chr.16: 58114036-58114036 on Build GRCh38
rs773105637 Chr.16: 58114052-58114052 on Build GRCh38
rs967249307 Chr.16: 58114007-58114007 on Build GRCh38
rs768915808 Chr.16: 58114023-58114023 on Build GRCh38
rs760225553 Chr.16: 58113961-58113961 on Build GRCh38
rs1021356739 Chr.16: 58113984-58113984 on Build GRCh38
rs976080523 Chr.16: 58114099-58114099 on Build GRCh38
rs534080716 Chr.16: 58114087-58114087 on Build GRCh38
rs755996604 Chr.16: 58114009-58114009 on Build GRCh38
rs112009407 Chr.16: 58114120-58114120 on Build GRCh38
rs369929997 Chr.16: 58113935-58113935 on Build GRCh38
rs894107905 Chr.16: 58113964-58113964 on Build GRCh38
rs1010946796 Chr.16: 58113969-58113969 on Build GRCh38
rs528581008 Chr.16: 58114062-58114062 on Build GRCh38
rs1029730061 Chr.16: 58114016-58114016 on Build GRCh38
rs745820488 Chr.16: 58114005-58114005 on Build GRCh38
rs752303043 Chr.16: 58113979-58113981 on Build GRCh38
rs573649492 Chr.16: 58114109-58114109 on Build GRCh38
rs748242297 Chr.16: 58114039-58114039 on Build GRCh38
rs369274875 Chr.16: 58113956-58113956 on Build GRCh38
rs751090482 Chr.16: 58113994-58113994 on Build GRCh38
rs772194076 Chr.16: 58114045-58114045 on Build GRCh38
rs372763790 Chr.16: 58113953-58113953 on Build GRCh38
rs146142113 Chr.16: 58113928-58113928 on Build GRCh38
rs780502574 Chr.16: 58114003-58114003 on Build GRCh38
rs1032815503 Chr.16: 58114051-58114051 on Build GRCh38
rs371099110 Chr.16: 58114080-58114080 on Build GRCh38
rs763784541 Chr.16: 58113986-58113986 on Build GRCh38
rs200765210 Chr.16: 58114002-58114002 on Build GRCh38
rs893689710 Chr.16: 58113954-58113954 on Build GRCh38
rs35368178 Chr.16: 58113948-58113948 on Build GRCh38
rs918902468 Chr.16: 58114119-58114119 on Build GRCh38

View Full Product Details