Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 82389667-82389922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL25
Entrez Gene ID
84190
Gene Name
methyltransferase like 25
Gene Aliases
C12orf26
UniGene
Hs.506222
Gene Chromosome Location
Chr.12: 82358497-82479239 on Build GRCh38
Transcripts
NM_001319675.1 NM_032230.2 XM_005269186.4 XM_011538827.2 XM_011538828.2 XM_011538829.2 XM_017020020.1 XM_017020021.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1993789 c.528G>A p.K176K Chr.12: 82389919-82389919 on Build GRCh38
1686681 c.425-1G>A p.? Chr.12: 82389815-82389815 on Build GRCh38
6553566 c.460A>T p.K154* Chr.12: 82389851-82389851 on Build GRCh38
1993788 c.508G>A p.A170T Chr.12: 82389899-82389899 on Build GRCh38
3813113 c.490G>C p.E164Q Chr.12: 82389881-82389881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774942705 Chr.12: 82389794-82389797 on Build GRCh38
rs761670272 Chr.12: 82389870-82389870 on Build GRCh38
rs527681441 Chr.12: 82389693-82389693 on Build GRCh38
rs556706891 Chr.12: 82389768-82389768 on Build GRCh38
rs763466100 Chr.12: 82389801-82389805 on Build GRCh38
rs549105540 Chr.12: 82389699-82389699 on Build GRCh38
rs748601236 Chr.12: 82389803-82389803 on Build GRCh38
rs753320978 Chr.12: 82389801-82389802 on Build GRCh38
rs780966087 Chr.12: 82389790-82389790 on Build GRCh38
rs1040676479 Chr.12: 82389892-82389892 on Build GRCh38
rs369465475 Chr.12: 82389884-82389884 on Build GRCh38
rs10778908 Chr.12: 82389889-82389889 on Build GRCh38
rs764880199 Chr.12: 82389872-82389872 on Build GRCh38
rs763312726 Chr.12: 82389831-82389831 on Build GRCh38
rs752857168 Chr.12: 82389859-82389859 on Build GRCh38
rs201870339 Chr.12: 82389875-82389875 on Build GRCh38
rs746695529 Chr.12: 82389779-82389779 on Build GRCh38
rs774374204 Chr.12: 82389837-82389837 on Build GRCh38
rs567380040 Chr.12: 82389712-82389712 on Build GRCh38
rs759588985 Chr.12: 82389840-82389840 on Build GRCh38
rs535270097 Chr.12: 82389742-82389742 on Build GRCh38
rs200647668 Chr.12: 82389816-82389816 on Build GRCh38
rs983322165 Chr.12: 82389826-82389826 on Build GRCh38
rs767784544 Chr.12: 82389849-82389849 on Build GRCh38
rs373688429 Chr.12: 82389832-82389832 on Build GRCh38
rs887121856 Chr.12: 82389813-82389813 on Build GRCh38
rs373003458 Chr.12: 82389891-82389891 on Build GRCh38
rs760266052 Chr.12: 82389798-82389799 on Build GRCh38
rs369502472 Chr.12: 82389811-82389811 on Build GRCh38
rs768460013 Chr.12: 82389781-82389781 on Build GRCh38

View Full Product Details