Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 127589957-127590218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARF5
Entrez Gene ID
381
Gene Name
ADP ribosylation factor 5
Gene Aliases
-
UniGene
Hs.653202
Gene Chromosome Location
Chr.7: 127588352-127591705 on Build GRCh38
Transcripts
NM_001662.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3632951 c.259-1G>A p.? Chr.7: 127590065-127590065 on Build GRCh38
5567118 c.290G>A p.R97Q Chr.7: 127590097-127590097 on Build GRCh38
5484893 c.296G>T p.R99L Chr.7: 127590103-127590103 on Build GRCh38
78870 c.292G>T p.E98* Chr.7: 127590099-127590099 on Build GRCh38
1085148 c.304G>T p.E102* Chr.7: 127590111-127590111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781588986 Chr.7: 127590117-127590117 on Build GRCh38
rs111443600 Chr.7: 127590186-127590186 on Build GRCh38
rs771445967 Chr.7: 127590114-127590114 on Build GRCh38
rs904485539 Chr.7: 127590055-127590055 on Build GRCh38
rs746280604 Chr.7: 127590006-127590006 on Build GRCh38
rs747475395 Chr.7: 127590113-127590113 on Build GRCh38
rs529540985 Chr.7: 127590010-127590010 on Build GRCh38
rs748365534 Chr.7: 127590042-127590045 on Build GRCh38
rs747116387 Chr.7: 127590162-127590162 on Build GRCh38
rs367582686 Chr.7: 127590173-127590173 on Build GRCh38
rs1049457227 Chr.7: 127590153-127590153 on Build GRCh38
rs772402404 Chr.7: 127590045-127590046 on Build GRCh38
rs753649005 Chr.7: 127590040-127590040 on Build GRCh38
rs367999369 Chr.7: 127590096-127590096 on Build GRCh38
rs371970454 Chr.7: 127590158-127590158 on Build GRCh38
rs777834274 Chr.7: 127590104-127590104 on Build GRCh38
rs754771745 Chr.7: 127590071-127590071 on Build GRCh38
rs965365418 Chr.7: 127590193-127590193 on Build GRCh38
rs374077814 Chr.7: 127590182-127590182 on Build GRCh38
rs766043154 Chr.7: 127590039-127590039 on Build GRCh38
rs1036658847 Chr.7: 127590187-127590187 on Build GRCh38
rs772875588 Chr.7: 127590185-127590185 on Build GRCh38
rs376627845 Chr.7: 127590160-127590160 on Build GRCh38
rs773130401 Chr.7: 127590046-127590049 on Build GRCh38
rs144009854 Chr.7: 127590097-127590097 on Build GRCh38
rs544463128 Chr.7: 127590172-127590172 on Build GRCh38
rs760689131 Chr.7: 127590030-127590030 on Build GRCh38
rs113460265 Chr.7: 127590138-127590138 on Build GRCh38
rs977424794 Chr.7: 127590045-127590045 on Build GRCh38
rs774086281 Chr.7: 127590165-127590165 on Build GRCh38
rs956252786 Chr.7: 127590147-127590147 on Build GRCh38
rs750315492 Chr.7: 127590017-127590017 on Build GRCh38
rs149540717 Chr.7: 127590080-127590080 on Build GRCh38

View Full Product Details