Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 49155756-49156027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf128
Entrez Gene ID
170371
Gene Name
chromosome 10 open reading frame 128
Gene Aliases
-
UniGene
Hs.385493
Gene Chromosome Location
Chr.10: 49154724-49188580 on Build GRCh38
Transcripts
NM_001010863.2 NM_001288740.1 NM_001288741.1 NM_001288742.1 NM_001288743.1 XM_011539379.1 XM_011539380.1 XM_011539383.2 XM_011539384.1 XM_011539385.1 XM_017015792.1 XM_017015793.1 XM_017015794.1 XM_017015797.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6006018 c.294-2A>T p.? Chr.10: 49155911-49155911 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375388588 Chr.10: 49155896-49155896 on Build GRCh38
rs943119062 Chr.10: 49155948-49155948 on Build GRCh38
rs775429071 Chr.10: 49155797-49155797 on Build GRCh38
rs755676951 Chr.10: 49155871-49155871 on Build GRCh38
rs759372169 Chr.10: 49155952-49155952 on Build GRCh38
rs919187716 Chr.10: 49155893-49155893 on Build GRCh38
rs141518641 Chr.10: 49155851-49155851 on Build GRCh38
rs535141089 Chr.10: 49155971-49155971 on Build GRCh38
rs967531518 Chr.10: 49155817-49155817 on Build GRCh38
rs903445737 Chr.10: 49155977-49155977 on Build GRCh38
rs971936185 Chr.10: 49155887-49155887 on Build GRCh38
rs760824370 Chr.10: 49155908-49155908 on Build GRCh38
rs761956901 Chr.10: 49155922-49155922 on Build GRCh38
rs776189254 Chr.10: 49155900-49155900 on Build GRCh38
rs747632689 Chr.10: 49155864-49155864 on Build GRCh38
rs576934497 Chr.10: 49155972-49155972 on Build GRCh38
rs566867300 Chr.10: 49155967-49155967 on Build GRCh38
rs1033211802 Chr.10: 49155844-49155844 on Build GRCh38
rs143639349 Chr.10: 49155880-49155880 on Build GRCh38
rs771268325 Chr.10: 49155876-49155876 on Build GRCh38
rs75050051 Chr.10: 49155796-49155796 on Build GRCh38
rs746344552 Chr.10: 49155890-49155890 on Build GRCh38
rs776846893 Chr.10: 49155909-49155909 on Build GRCh38
rs751459951 Chr.10: 49155854-49155854 on Build GRCh38
rs910409393 Chr.10: 49155944-49155944 on Build GRCh38
rs932076954 Chr.10: 49155982-49155982 on Build GRCh38
rs752696695 Chr.10: 49155961-49155961 on Build GRCh38
rs780644410 Chr.10: 49155856-49155856 on Build GRCh38
rs766151854 Chr.10: 49155929-49155929 on Build GRCh38
rs536631405 Chr.10: 49155852-49155852 on Build GRCh38
rs767425030 Chr.10: 49155957-49155957 on Build GRCh38
rs901480378 Chr.10: 49155953-49155953 on Build GRCh38
rs912511765 Chr.10: 49155939-49155939 on Build GRCh38
rs201099327 Chr.10: 49155855-49155855 on Build GRCh38
rs371134595 Chr.10: 49155873-49155873 on Build GRCh38
rs372417788 Chr.10: 49155966-49155966 on Build GRCh38
rs867710511 Chr.10: 49155915-49155915 on Build GRCh38
rs368506386 Chr.10: 49155942-49155942 on Build GRCh38
rs373193962 Chr.10: 49155837-49155837 on Build GRCh38
rs760879466 Chr.10: 49155836-49155836 on Build GRCh38
rs754171133 Chr.10: 49155841-49155841 on Build GRCh38
rs375626071 Chr.10: 49155872-49155872 on Build GRCh38

View Full Product Details