Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7090051-7090290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1R
Entrez Gene ID
715
Gene Name
complement C1r subcomponent
Gene Aliases
-
UniGene
Hs.524224
Gene Chromosome Location
Chr.12: 7080209-7092447 on Build GRCh38
Transcripts
NM_001733.4 XM_017019915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
943311 c.356T>G p.F119C Chr.12: 7090166-7090166 on Build GRCh38
5789836 c.466+3G>A p.? Chr.12: 7090053-7090053 on Build GRCh38
4637695 c.338C>A p.P113H Chr.12: 7090184-7090184 on Build GRCh38
3465076 c.405C>T p.S135S Chr.12: 7090117-7090117 on Build GRCh38
943312 c.323C>A p.S108Y Chr.12: 7090199-7090199 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758802004 Chr.12: 7090249-7090249 on Build GRCh38
rs768596165 Chr.12: 7090098-7090098 on Build GRCh38
rs772213361 Chr.12: 7090078-7090078 on Build GRCh38
rs747528717 Chr.12: 7090110-7090110 on Build GRCh38
rs780301260 Chr.12: 7090163-7090163 on Build GRCh38
rs766205007 Chr.12: 7090119-7090119 on Build GRCh38
rs746101398 Chr.12: 7090076-7090076 on Build GRCh38
rs747997099 Chr.12: 7090101-7090101 on Build GRCh38
rs150953301 Chr.12: 7090114-7090114 on Build GRCh38
rs759058268 Chr.12: 7090221-7090221 on Build GRCh38
rs751803218 Chr.12: 7090257-7090257 on Build GRCh38
rs769654256 Chr.12: 7090105-7090105 on Build GRCh38
rs4600291 Chr.12: 7090267-7090267 on Build GRCh38
rs139531404 Chr.12: 7090144-7090144 on Build GRCh38
rs769553294 Chr.12: 7090190-7090190 on Build GRCh38
rs776176820 Chr.12: 7090123-7090123 on Build GRCh38
rs149665060 Chr.12: 7090146-7090146 on Build GRCh38
rs750515595 Chr.12: 7090100-7090100 on Build GRCh38
rs763828463 Chr.12: 7090265-7090265 on Build GRCh38
rs765720381 Chr.12: 7090239-7090239 on Build GRCh38
rs781084847 Chr.12: 7090180-7090180 on Build GRCh38
rs768142600 Chr.12: 7090219-7090219 on Build GRCh38
rs747201064 Chr.12: 7090170-7090170 on Build GRCh38
rs750327032 Chr.12: 7090133-7090133 on Build GRCh38
rs760060680 Chr.12: 7090177-7090178 on Build GRCh38
rs368415263 Chr.12: 7090194-7090194 on Build GRCh38
rs774190714 Chr.12: 7090218-7090218 on Build GRCh38
rs994317175 Chr.12: 7090245-7090245 on Build GRCh38
rs374748362 Chr.12: 7090153-7090153 on Build GRCh38
rs779572553 Chr.12: 7090225-7090225 on Build GRCh38
rs766689135 Chr.12: 7090253-7090253 on Build GRCh38
rs766909412 Chr.12: 7090141-7090141 on Build GRCh38
rs750849894 Chr.12: 7090155-7090155 on Build GRCh38
rs748130929 Chr.12: 7090181-7090181 on Build GRCh38
rs758746826 Chr.12: 7090158-7090158 on Build GRCh38
rs780249848 Chr.12: 7090085-7090085 on Build GRCh38
rs760225012 Chr.12: 7090231-7090231 on Build GRCh38
rs376041687 Chr.12: 7090199-7090199 on Build GRCh38
rs142544799 Chr.12: 7090113-7090113 on Build GRCh38
rs774793614 Chr.12: 7090230-7090230 on Build GRCh38
rs755115134 Chr.12: 7090174-7090174 on Build GRCh38
rs200185988 Chr.12: 7090203-7090203 on Build GRCh38
rs758237909 Chr.12: 7090150-7090150 on Build GRCh38
rs747147957 Chr.12: 7090088-7090088 on Build GRCh38
rs371672761 Chr.12: 7090243-7090243 on Build GRCh38
rs371144067 Chr.12: 7090149-7090149 on Build GRCh38

View Full Product Details