Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2280693-2280958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf35
Entrez Gene ID
374872
Gene Name
chromosome 19 open reading frame 35
Gene Aliases
-
UniGene
Hs.511803
Gene Chromosome Location
Chr.19: 2274632-2282182 on Build GRCh38
Transcripts
NM_198532.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4921034 c.34G>A p.E12K Chr.19: 2280898-2280898 on Build GRCh38
4076378 c.68C>T p.T23M Chr.19: 2280864-2280864 on Build GRCh38
6406881 c.25G>T p.E9* Chr.19: 2280907-2280907 on Build GRCh38
4667162 c.74G>A p.S25N Chr.19: 2280858-2280858 on Build GRCh38
3378672 c.14A>G p.E5G Chr.19: 2280918-2280918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575727730 Chr.19: 2280829-2280829 on Build GRCh38
rs753314460 Chr.19: 2280795-2280795 on Build GRCh38
rs774572909 Chr.19: 2280825-2280825 on Build GRCh38
rs761268907 Chr.19: 2280843-2280843 on Build GRCh38
rs147999963 Chr.19: 2280920-2280920 on Build GRCh38
rs993825157 Chr.19: 2280748-2280748 on Build GRCh38
rs780108572 Chr.19: 2280854-2280854 on Build GRCh38
rs371625189 Chr.19: 2280832-2280832 on Build GRCh38
rs1031817279 Chr.19: 2280794-2280794 on Build GRCh38
rs368167267 Chr.19: 2280818-2280818 on Build GRCh38
rs748220415 Chr.19: 2280813-2280813 on Build GRCh38
rs897718535 Chr.19: 2280728-2280728 on Build GRCh38
rs374254097 Chr.19: 2280921-2280921 on Build GRCh38
rs763374432 Chr.19: 2280817-2280817 on Build GRCh38
rs749444997 Chr.19: 2280866-2280866 on Build GRCh38
rs781519940 Chr.19: 2280863-2280863 on Build GRCh38
rs780551178 Chr.19: 2280899-2280899 on Build GRCh38
rs771063954 Chr.19: 2280914-2280914 on Build GRCh38
rs535290401 Chr.19: 2280764-2280764 on Build GRCh38
rs367577938 Chr.19: 2280850-2280850 on Build GRCh38
rs545718575 Chr.19: 2280765-2280765 on Build GRCh38
rs767660808 Chr.19: 2280774-2280792 on Build GRCh38
rs915324573 Chr.19: 2280744-2280744 on Build GRCh38
rs775737691 Chr.19: 2280742-2280742 on Build GRCh38
rs763964908 Chr.19: 2280752-2280753 on Build GRCh38
rs980498056 Chr.19: 2280819-2280819 on Build GRCh38
rs866341495 Chr.19: 2280749-2280749 on Build GRCh38
rs140660078 Chr.19: 2280904-2280904 on Build GRCh38
rs750314161 Chr.19: 2280851-2280851 on Build GRCh38
rs55662626 Chr.19: 2280886-2280886 on Build GRCh38
rs763801334 Chr.19: 2280793-2280793 on Build GRCh38
rs1056325778 Chr.19: 2280719-2280719 on Build GRCh38
rs756278154 Chr.19: 2280910-2280910 on Build GRCh38
rs375740830 Chr.19: 2280869-2280869 on Build GRCh38
rs757246222 Chr.19: 2280772-2280772 on Build GRCh38
rs926332941 Chr.19: 2280828-2280828 on Build GRCh38
rs764995528 Chr.19: 2280893-2280893 on Build GRCh38
rs143764794 Chr.19: 2280915-2280915 on Build GRCh38
rs763328728 Chr.19: 2280774-2280774 on Build GRCh38
rs371294668 Chr.19: 2280803-2280803 on Build GRCh38
rs758253320 Chr.19: 2280852-2280852 on Build GRCh38
rs750589842 Chr.19: 2280778-2280797 on Build GRCh38
rs12460518 Chr.19: 2280797-2280797 on Build GRCh38
rs868519925 Chr.19: 2280930-2280930 on Build GRCh38
rs761464605 Chr.19: 2280892-2280892 on Build GRCh38
rs372360199 Chr.19: 2280899-2280899 on Build GRCh38
rs1023556509 Chr.19: 2280773-2280773 on Build GRCh38
rs930536013 Chr.19: 2280917-2280917 on Build GRCh38
rs776194446 Chr.19: 2280891-2280891 on Build GRCh38
rs770020666 Chr.19: 2280814-2280814 on Build GRCh38
rs369840674 Chr.19: 2280909-2280909 on Build GRCh38
rs773645747 Chr.19: 2280767-2280767 on Build GRCh38
rs768794996 Chr.19: 2280762-2280762 on Build GRCh38
rs766208559 Chr.19: 2280900-2280900 on Build GRCh38
rs756809251 Chr.19: 2280796-2280796 on Build GRCh38
rs202241723 Chr.19: 2280758-2280777 on Build GRCh38
rs374242436 Chr.19: 2280804-2280804 on Build GRCh38
rs759932346 Chr.19: 2280826-2280826 on Build GRCh38
rs747198079 Chr.19: 2280746-2280746 on Build GRCh38
rs113248494 Chr.19: 2280784-2280784 on Build GRCh38
rs751372660 Chr.19: 2280857-2280857 on Build GRCh38
rs752328649 Chr.19: 2280783-2280783 on Build GRCh38
rs749305194 Chr.19: 2280911-2280911 on Build GRCh38
rs756444829 Chr.19: 2280892-2280893 on Build GRCh38
rs776840194 Chr.19: 2280763-2280763 on Build GRCh38
rs980873573 Chr.19: 2280898-2280898 on Build GRCh38
rs745820667 Chr.19: 2280801-2280801 on Build GRCh38
rs10420010 Chr.19: 2280777-2280777 on Build GRCh38
rs774970677 Chr.19: 2280875-2280875 on Build GRCh38
rs772456363 Chr.19: 2280887-2280887 on Build GRCh38
rs976191098 Chr.19: 2280792-2280792 on Build GRCh38
rs377061660 Chr.19: 2280902-2280902 on Build GRCh38
rs141253643 Chr.19: 2280864-2280864 on Build GRCh38
rs777036390 Chr.19: 2280807-2280807 on Build GRCh38

View Full Product Details