Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 26496331-26496566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBB1IP
Entrez Gene ID
54518
Gene Name
amyloid beta precursor protein binding family B member 1 interacting protein
Gene Aliases
INAG1, PREL1, RARP1, RIAM
UniGene
Hs.310421
Gene Chromosome Location
Chr.10: 26438324-26567879 on Build GRCh38
Transcripts
NM_019043.3 XM_006717451.2 XM_011519514.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5130164 c.100C>G p.P34A Chr.10: 26496331-26496331 on Build GRCh38
3436873 c.160G>A p.E54K Chr.10: 26496391-26496391 on Build GRCh38
3436874 c.160G>A p.E54K Chr.10: 26496391-26496391 on Build GRCh38
4984512 c.116C>A p.P39H Chr.10: 26496347-26496347 on Build GRCh38
4984511 c.116C>A p.P39H Chr.10: 26496347-26496347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970293578 Chr.10: 26496496-26496496 on Build GRCh38
rs781737065 Chr.10: 26496351-26496351 on Build GRCh38
rs933310739 Chr.10: 26496383-26496383 on Build GRCh38
rs987877849 Chr.10: 26496452-26496452 on Build GRCh38
rs980296190 Chr.10: 26496504-26496504 on Build GRCh38
rs921855367 Chr.10: 26496354-26496354 on Build GRCh38
rs955930488 Chr.10: 26496463-26496465 on Build GRCh38
rs914186079 Chr.10: 26496489-26496489 on Build GRCh38
rs907911480 Chr.10: 26496465-26496465 on Build GRCh38
rs770437464 Chr.10: 26496375-26496375 on Build GRCh38
rs771819264 Chr.10: 26496441-26496441 on Build GRCh38
rs745583284 Chr.10: 26496394-26496394 on Build GRCh38
rs1028172656 Chr.10: 26496353-26496353 on Build GRCh38
rs368860973 Chr.10: 26496505-26496505 on Build GRCh38
rs748677908 Chr.10: 26496358-26496358 on Build GRCh38
rs940755062 Chr.10: 26496487-26496487 on Build GRCh38
rs773977870 Chr.10: 26496380-26496380 on Build GRCh38
rs373211324 Chr.10: 26496382-26496382 on Build GRCh38

View Full Product Details