Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 98089302-98089571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH5
Entrez Gene ID
203111
Gene Name
glutamate rich 5
Gene Aliases
C8orf47
UniGene
Hs.171455
Gene Chromosome Location
Chr.8: 98064522-98093610 on Build GRCh38
Transcripts
NM_001170806.1 NM_173549.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3902331 c.423G>C p.E141D Chr.8: 98089440-98089440 on Build GRCh38
1102868 c.436A>C p.N146H Chr.8: 98089453-98089453 on Build GRCh38
6181672 c.366G>A p.E122E Chr.8: 98089383-98089383 on Build GRCh38
5625872 c.344C>T p.P115L Chr.8: 98089361-98089361 on Build GRCh38
3367447 c.425C>T p.S142F Chr.8: 98089442-98089442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776589874 Chr.8: 98089545-98089545 on Build GRCh38
rs761432288 Chr.8: 98089533-98089533 on Build GRCh38
rs748202835 Chr.8: 98089324-98089324 on Build GRCh38
rs770690639 Chr.8: 98089349-98089349 on Build GRCh38
rs774167601 Chr.8: 98089354-98089354 on Build GRCh38
rs933281427 Chr.8: 98089395-98089395 on Build GRCh38
rs756204132 Chr.8: 98089432-98089432 on Build GRCh38
rs752928685 Chr.8: 98089427-98089427 on Build GRCh38
rs751924915 Chr.8: 98089398-98089398 on Build GRCh38
rs555991242 Chr.8: 98089371-98089371 on Build GRCh38
rs749214826 Chr.8: 98089437-98089437 on Build GRCh38
rs374440486 Chr.8: 98089345-98089345 on Build GRCh38
rs149544768 Chr.8: 98089438-98089438 on Build GRCh38
rs763572719 Chr.8: 98089384-98089384 on Build GRCh38
rs146451719 Chr.8: 98089361-98089361 on Build GRCh38
rs138721529 Chr.8: 98089362-98089362 on Build GRCh38
rs761689088 Chr.8: 98089399-98089400 on Build GRCh38
rs867556143 Chr.8: 98089489-98089489 on Build GRCh38
rs199854704 Chr.8: 98089493-98089493 on Build GRCh38
rs1049093101 Chr.8: 98089471-98089471 on Build GRCh38
rs539687893 Chr.8: 98089507-98089507 on Build GRCh38
rs767024963 Chr.8: 98089388-98089388 on Build GRCh38
rs778840856 Chr.8: 98089485-98089485 on Build GRCh38
rs370646763 Chr.8: 98089545-98089545 on Build GRCh38
rs777764848 Chr.8: 98089433-98089433 on Build GRCh38
rs773195953 Chr.8: 98089337-98089337 on Build GRCh38
rs774471187 Chr.8: 98089328-98089329 on Build GRCh38
rs35339824 Chr.8: 98089326-98089326 on Build GRCh38
rs111454971 Chr.8: 98089543-98089543 on Build GRCh38
rs781495538 Chr.8: 98089416-98089416 on Build GRCh38
rs771923338 Chr.8: 98089494-98089494 on Build GRCh38
rs750923722 Chr.8: 98089387-98089387 on Build GRCh38
rs371062621 Chr.8: 98089333-98089333 on Build GRCh38
rs760230946 Chr.8: 98089373-98089373 on Build GRCh38
rs144156466 Chr.8: 98089513-98089513 on Build GRCh38
rs766434641 Chr.8: 98089504-98089506 on Build GRCh38
rs769429935 Chr.8: 98089518-98089518 on Build GRCh38
rs755408926 Chr.8: 98089399-98089399 on Build GRCh38

View Full Product Details