Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 76459875-76460146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMSY
Entrez Gene ID
56946
Gene Name
EMSY, BRCA2 interacting transcriptional repressor
Gene Aliases
C11orf30, GL002
UniGene
Hs.352588
Gene Chromosome Location
Chr.11: 76444928-76552901 on Build GRCh38
Transcripts
NM_001300942.1 NM_001300943.1 NM_001300944.1 NM_020193.4 XM_005274112.4 XM_006718637.3 XM_006718638.3 XM_006718639.3 XM_011545173.2 XM_011545174.2 XM_011545175.2 XM_011545177.2 XM_011545178.2 XM_011545179.2 XM_017018021.1 XM_017018022.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3736049 c.510A>C p.R170S Chr.11: 76460024-76460024 on Build GRCh38
4666597 c.487A>G p.I163V Chr.11: 76460001-76460001 on Build GRCh38
5575499 c.571A>T p.S191C Chr.11: 76460085-76460085 on Build GRCh38
5017729 c.449C>T p.T150M Chr.11: 76459963-76459963 on Build GRCh38
231869 c.511C>T p.P171S Chr.11: 76460025-76460025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556789767 Chr.11: 76460114-76460114 on Build GRCh38
rs756978004 Chr.11: 76460017-76460017 on Build GRCh38
rs1024566153 Chr.11: 76459932-76459932 on Build GRCh38
rs773721122 Chr.11: 76459897-76459897 on Build GRCh38
rs751201336 Chr.11: 76460010-76460010 on Build GRCh38
rs766093465 Chr.11: 76459958-76459958 on Build GRCh38
rs773370981 Chr.11: 76459930-76459930 on Build GRCh38
rs780581672 Chr.11: 76459896-76459896 on Build GRCh38
rs141598201 Chr.11: 76459999-76459999 on Build GRCh38
rs372186650 Chr.11: 76459963-76459963 on Build GRCh38
rs765136304 Chr.11: 76459982-76459982 on Build GRCh38
rs762446624 Chr.11: 76459962-76459962 on Build GRCh38
rs377074594 Chr.11: 76459989-76459989 on Build GRCh38
rs970590801 Chr.11: 76459965-76459965 on Build GRCh38
rs913275147 Chr.11: 76460103-76460103 on Build GRCh38
rs980313601 Chr.11: 76459990-76459990 on Build GRCh38
rs1052084066 Chr.11: 76460111-76460111 on Build GRCh38
rs755474883 Chr.11: 76460020-76460020 on Build GRCh38
rs369586337 Chr.11: 76460100-76460100 on Build GRCh38
rs763722061 Chr.11: 76460008-76460008 on Build GRCh38
rs760898724 Chr.11: 76459951-76459951 on Build GRCh38
rs376653441 Chr.11: 76459922-76459922 on Build GRCh38
rs373792798 Chr.11: 76460107-76460107 on Build GRCh38
rs777832208 Chr.11: 76460038-76460038 on Build GRCh38
rs540076045 Chr.11: 76460057-76460057 on Build GRCh38
rs369145611 Chr.11: 76459923-76459923 on Build GRCh38
rs150715503 Chr.11: 76460121-76460121 on Build GRCh38
rs769113365 Chr.11: 76459911-76459911 on Build GRCh38
rs780565267 Chr.11: 76460018-76460018 on Build GRCh38
rs911816843 Chr.11: 76460113-76460113 on Build GRCh38
rs759504010 Chr.11: 76459964-76459964 on Build GRCh38
rs749676543 Chr.11: 76460019-76460019 on Build GRCh38
rs771985129 Chr.11: 76460024-76460024 on Build GRCh38
rs748707730 Chr.11: 76460023-76460023 on Build GRCh38
rs866533903 Chr.11: 76460021-76460024 on Build GRCh38
rs749749282 Chr.11: 76459908-76459908 on Build GRCh38

View Full Product Details