Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 51607438-51607691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP9A
Entrez Gene ID
10079
Gene Name
ATPase phospholipid transporting 9A (putative)
Gene Aliases
ATPIIA
UniGene
Hs.649234
Gene Chromosome Location
Chr.20: 51596775-51768411 on Build GRCh38
Transcripts
NM_006045.2 XM_011528481.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3972608 c.2778A>G p.I926M Chr.20: 51607552-51607552 on Build GRCh38
6356745 c.2746G>A p.G916R Chr.20: 51607584-51607584 on Build GRCh38
5657831 c.2746-1G>T p.? Chr.20: 51607585-51607585 on Build GRCh38
69773 c.2770T>G p.F924V Chr.20: 51607560-51607560 on Build GRCh38
6871098 c.2772C>T p.F924F Chr.20: 51607558-51607558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534785099 Chr.20: 51607638-51607638 on Build GRCh38
rs778403529 Chr.20: 51607480-51607480 on Build GRCh38
rs767539776 Chr.20: 51607580-51607580 on Build GRCh38
rs755232436 Chr.20: 51607633-51607633 on Build GRCh38
rs750753976 Chr.20: 51607622-51607622 on Build GRCh38
rs758906132 Chr.20: 51607486-51607486 on Build GRCh38
rs200339048 Chr.20: 51607492-51607492 on Build GRCh38
rs900388206 Chr.20: 51607656-51607656 on Build GRCh38
rs553122035 Chr.20: 51607655-51607655 on Build GRCh38
rs762319389 Chr.20: 51607466-51607467 on Build GRCh38
rs954538668 Chr.20: 51607666-51607666 on Build GRCh38
rs370337954 Chr.20: 51607634-51607634 on Build GRCh38
rs756918151 Chr.20: 51607617-51607617 on Build GRCh38
rs533169778 Chr.20: 51607532-51607532 on Build GRCh38
rs745436564 Chr.20: 51607484-51607484 on Build GRCh38
rs1055181385 Chr.20: 51607593-51607593 on Build GRCh38
rs567449180 Chr.20: 51607623-51607623 on Build GRCh38
rs763164950 Chr.20: 51607576-51607576 on Build GRCh38
rs753370622 Chr.20: 51607615-51607615 on Build GRCh38
rs948824818 Chr.20: 51607474-51607474 on Build GRCh38
rs2273089 Chr.20: 51607622-51607622 on Build GRCh38
rs748518698 Chr.20: 51607636-51607636 on Build GRCh38
rs551441379 Chr.20: 51607608-51607608 on Build GRCh38
rs3818004 Chr.20: 51607667-51607667 on Build GRCh38
rs955213522 Chr.20: 51607664-51607664 on Build GRCh38
rs140801520 Chr.20: 51607501-51607501 on Build GRCh38
rs997010696 Chr.20: 51607625-51607625 on Build GRCh38
rs763951428 Chr.20: 51607614-51607614 on Build GRCh38
rs376369640 Chr.20: 51607509-51607509 on Build GRCh38
rs116400327 Chr.20: 51607665-51607665 on Build GRCh38
rs372002496 Chr.20: 51607495-51607495 on Build GRCh38
rs147701100 Chr.20: 51607472-51607472 on Build GRCh38
rs767628076 Chr.20: 51607610-51607610 on Build GRCh38
rs756840211 Chr.20: 51607494-51607494 on Build GRCh38
rs745629271 Chr.20: 51607496-51607496 on Build GRCh38
rs773412527 Chr.20: 51607535-51607535 on Build GRCh38
rs747583567 Chr.20: 51607628-51607628 on Build GRCh38
rs569981759 Chr.20: 51607620-51607620 on Build GRCh38
rs775589680 Chr.20: 51607581-51607581 on Build GRCh38
rs536937443 Chr.20: 51607621-51607621 on Build GRCh38
rs767790273 Chr.20: 51607591-51607592 on Build GRCh38

View Full Product Details