Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 42270867-42271126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCG1
Entrez Gene ID
9619
Gene Name
ATP binding cassette subfamily G member 1
Gene Aliases
ABC8, WHITE1
UniGene
Hs.124649
Gene Chromosome Location
Chr.21: 42199689-42304389 on Build GRCh38
Transcripts
NM_004915.3 NM_016818.2 NM_207174.1 NM_207627.1 NM_207628.1 NM_207629.1 XM_011529806.1 XM_011529807.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4959509 c.338G>A p.G113D Chr.21: 42271115-42271115 on Build GRCh38
5644292 c.309G>T p.L103L Chr.21: 42271086-42271086 on Build GRCh38
5612084 c.725-2A>T p.? Chr.21: 42271068-42271068 on Build GRCh38
6526303 c.725G>A p.G242E Chr.21: 42271070-42271070 on Build GRCh38
4419112 c.777G>A p.L259L Chr.21: 42271122-42271122 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779637796 Chr.21: 42270954-42270954 on Build GRCh38
rs762828210 Chr.21: 42271099-42271099 on Build GRCh38
rs781275773 Chr.21: 42271022-42271022 on Build GRCh38
rs764034522 Chr.21: 42271101-42271101 on Build GRCh38
rs17114604 Chr.21: 42271029-42271029 on Build GRCh38
rs375599514 Chr.21: 42271080-42271080 on Build GRCh38
rs78923761 Chr.21: 42270902-42270902 on Build GRCh38
rs757443901 Chr.21: 42271021-42271021 on Build GRCh38
rs906858480 Chr.21: 42271026-42271026 on Build GRCh38
rs115474443 Chr.21: 42270956-42270956 on Build GRCh38
rs758064280 Chr.21: 42270935-42270935 on Build GRCh38
rs191666990 Chr.21: 42271098-42271098 on Build GRCh38
rs769315931 Chr.21: 42271041-42271041 on Build GRCh38
rs746511159 Chr.21: 42271045-42271045 on Build GRCh38
rs772797947 Chr.21: 42271042-42271042 on Build GRCh38
rs868470577 Chr.21: 42270918-42270918 on Build GRCh38
rs759680276 Chr.21: 42271060-42271060 on Build GRCh38
rs527267427 Chr.21: 42271064-42271064 on Build GRCh38
rs991086924 Chr.21: 42270975-42270975 on Build GRCh38
rs769037111 Chr.21: 42271006-42271006 on Build GRCh38
rs958634423 Chr.21: 42270974-42270974 on Build GRCh38
rs1014783726 Chr.21: 42271052-42271052 on Build GRCh38
rs759360627 Chr.21: 42271067-42271067 on Build GRCh38
rs780687805 Chr.21: 42271033-42271033 on Build GRCh38
rs770546800 Chr.21: 42271062-42271062 on Build GRCh38
rs371020544 Chr.21: 42271036-42271036 on Build GRCh38
rs967690163 Chr.21: 42270928-42270928 on Build GRCh38
rs747367599 Chr.21: 42270972-42270972 on Build GRCh38
rs1006029529 Chr.21: 42271046-42271046 on Build GRCh38
rs375674037 Chr.21: 42271028-42271028 on Build GRCh38
rs140084573 Chr.21: 42270984-42270984 on Build GRCh38

View Full Product Details