Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109768718-109768961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM222A
Entrez Gene ID
84915
Gene Name
family with sequence similarity 222 member A
Gene Aliases
C12orf34
UniGene
Hs.661785
Gene Chromosome Location
Chr.12: 109714382-109770509 on Build GRCh38
Transcripts
NM_032829.2 XM_006719654.2 XM_017020055.1 NR_026661.2 NR_026662.2

Gene Symbol
FAM222A-AS1
Entrez Gene ID
84983
Gene Name
FAM222A antisense RNA 1
Gene Aliases
-
UniGene
Hs.675961
Gene Chromosome Location
Chr.12: 109734209-109773487 on Build GRCh38
Transcripts
NM_032829.2 XM_006719654.2 XM_017020055.1 NR_026661.2 NR_026662.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5599154 c.935C>T p.S312F Chr.12: 109768864-109768864 on Build GRCh38
4170364 c.807G>T p.L269F Chr.12: 109768736-109768736 on Build GRCh38
4038584 c.982C>T p.P328S Chr.12: 109768911-109768911 on Build GRCh38
6382499 c.1006C>T p.P336S Chr.12: 109768935-109768935 on Build GRCh38
6381050 c.855C>G p.Y285* Chr.12: 109768784-109768784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953716263 Chr.12: 109768884-109768884 on Build GRCh38
rs532999688 Chr.12: 109768764-109768764 on Build GRCh38
rs776744269 Chr.12: 109768845-109768845 on Build GRCh38
rs375805951 Chr.12: 109768763-109768763 on Build GRCh38
rs767676227 Chr.12: 109768760-109768760 on Build GRCh38
rs894377282 Chr.12: 109768840-109768840 on Build GRCh38
rs754643198 Chr.12: 109768885-109768885 on Build GRCh38
rs759692083 Chr.12: 109768762-109768775 on Build GRCh38
rs765564397 Chr.12: 109768786-109768786 on Build GRCh38
rs764600932 Chr.12: 109768772-109768772 on Build GRCh38
rs751947224 Chr.12: 109768940-109768940 on Build GRCh38
rs372883214 Chr.12: 109768879-109768879 on Build GRCh38
rs775633177 Chr.12: 109768826-109768826 on Build GRCh38
rs1028505911 Chr.12: 109768881-109768881 on Build GRCh38
rs768848391 Chr.12: 109768905-109768905 on Build GRCh38
rs751665127 Chr.12: 109768758-109768758 on Build GRCh38
rs750782496 Chr.12: 109768787-109768787 on Build GRCh38
rs756034504 Chr.12: 109768894-109768894 on Build GRCh38
rs749051896 Chr.12: 109768896-109768896 on Build GRCh38
rs763547106 Chr.12: 109768939-109768939 on Build GRCh38
rs77798629 Chr.12: 109768827-109768827 on Build GRCh38
rs143015847 Chr.12: 109768802-109768803 on Build GRCh38
rs369041230 Chr.12: 109768935-109768935 on Build GRCh38
rs550094496 Chr.12: 109768830-109768830 on Build GRCh38
rs376914566 Chr.12: 109768897-109768897 on Build GRCh38
rs780841125 Chr.12: 109768809-109768809 on Build GRCh38
rs372654452 Chr.12: 109768745-109768745 on Build GRCh38
rs770210812 Chr.12: 109768928-109768928 on Build GRCh38
rs562758512 Chr.12: 109768740-109768740 on Build GRCh38
rs567732574 Chr.12: 109768817-109768817 on Build GRCh38
rs139486750 Chr.12: 109768796-109768796 on Build GRCh38
rs200880090 Chr.12: 109768831-109768831 on Build GRCh38
rs201569829 Chr.12: 109768825-109768825 on Build GRCh38
rs922186655 Chr.12: 109768794-109768794 on Build GRCh38
rs542798024 Chr.12: 109768805-109768805 on Build GRCh38
rs759726487 Chr.12: 109768746-109768746 on Build GRCh38
rs369923455 Chr.12: 109768783-109768783 on Build GRCh38
rs527356091 Chr.12: 109768813-109768813 on Build GRCh38
rs201882014 Chr.12: 109768878-109768878 on Build GRCh38
rs911851788 Chr.12: 109768913-109768913 on Build GRCh38
rs779816573 Chr.12: 109768798-109768798 on Build GRCh38
rs750787358 Chr.12: 109768876-109768876 on Build GRCh38
rs749069255 Chr.12: 109768822-109768822 on Build GRCh38
rs61735678 Chr.12: 109768814-109768814 on Build GRCh38
rs538694450 Chr.12: 109768850-109768850 on Build GRCh38
rs776600173 Chr.12: 109768926-109768926 on Build GRCh38
rs780898328 Chr.12: 109768892-109768892 on Build GRCh38
rs61752601 Chr.12: 109768904-109768904 on Build GRCh38
rs746528053 Chr.12: 109768804-109768804 on Build GRCh38
rs754400731 Chr.12: 109768773-109768773 on Build GRCh38
rs763348389 Chr.12: 109768877-109768877 on Build GRCh38
rs142278927 Chr.12: 109768895-109768895 on Build GRCh38
rs565842202 Chr.12: 109768893-109768893 on Build GRCh38

View Full Product Details