Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 62819128-62819387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG4C
Entrez Gene ID
84938
Gene Name
autophagy related 4C cysteine peptidase
Gene Aliases
APG4-C, APG4C, AUTL1, AUTL3
UniGene
Hs.7353
Gene Chromosome Location
Chr.1: 62784106-62865270 on Build GRCh38
Transcripts
NM_032852.3 NM_178221.2 XM_005271288.2 XM_011542313.2 XM_017002600.1 XM_017002601.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
274073 c.666G>T p.K222N Chr.1: 62819276-62819276 on Build GRCh38
4425424 c.534G>C p.L178L Chr.1: 62819144-62819144 on Build GRCh38
6841204 c.574C>T p.R192* Chr.1: 62819184-62819184 on Build GRCh38
299664 c.586T>G p.Y196D Chr.1: 62819196-62819196 on Build GRCh38
1183911 c.575G>A p.R192Q Chr.1: 62819185-62819185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551760214 Chr.1: 62819215-62819216 on Build GRCh38
rs756440040 Chr.1: 62819337-62819337 on Build GRCh38
rs757256584 Chr.1: 62819193-62819194 on Build GRCh38
rs531433477 Chr.1: 62819284-62819284 on Build GRCh38
rs545667264 Chr.1: 62819200-62819200 on Build GRCh38
rs34243881 Chr.1: 62819217-62819218 on Build GRCh38
rs775506668 Chr.1: 62819183-62819183 on Build GRCh38
rs201262005 Chr.1: 62819227-62819227 on Build GRCh38
rs754002648 Chr.1: 62819168-62819168 on Build GRCh38
rs756893631 Chr.1: 62819315-62819315 on Build GRCh38
rs766187159 Chr.1: 62819197-62819197 on Build GRCh38
rs752726160 Chr.1: 62819210-62819210 on Build GRCh38
rs763638385 Chr.1: 62819314-62819314 on Build GRCh38
rs770237135 Chr.1: 62819261-62819261 on Build GRCh38
rs774527039 Chr.1: 62819188-62819188 on Build GRCh38
rs762009826 Chr.1: 62819192-62819192 on Build GRCh38
rs145971105 Chr.1: 62819267-62819267 on Build GRCh38
rs780500162 Chr.1: 62819230-62819230 on Build GRCh38
rs763043306 Chr.1: 62819185-62819185 on Build GRCh38
rs781552979 Chr.1: 62819253-62819253 on Build GRCh38
rs746326307 Chr.1: 62819173-62819173 on Build GRCh38
rs764208566 Chr.1: 62819186-62819186 on Build GRCh38
rs756872355 Chr.1: 62819249-62819249 on Build GRCh38
rs756041806 Chr.1: 62819153-62819153 on Build GRCh38
rs780464724 Chr.1: 62819358-62819358 on Build GRCh38
rs372214312 Chr.1: 62819230-62819230 on Build GRCh38
rs901516988 Chr.1: 62819184-62819184 on Build GRCh38
rs779345173 Chr.1: 62819211-62819212 on Build GRCh38
rs759226255 Chr.1: 62819299-62819299 on Build GRCh38
rs886253978 Chr.1: 62819208-62819208 on Build GRCh38
rs770206856 Chr.1: 62819179-62819179 on Build GRCh38
rs772191648 Chr.1: 62819278-62819278 on Build GRCh38
rs765300744 Chr.1: 62819206-62819206 on Build GRCh38
rs1016064966 Chr.1: 62819232-62819232 on Build GRCh38
rs774473913 Chr.1: 62819275-62819275 on Build GRCh38
rs754882812 Chr.1: 62819201-62819201 on Build GRCh38
rs769257399 Chr.1: 62819353-62819356 on Build GRCh38
rs768719940 Chr.1: 62819271-62819271 on Build GRCh38
rs113265845 Chr.1: 62819162-62819162 on Build GRCh38
rs780517769 Chr.1: 62819263-62819263 on Build GRCh38
rs752674789 Chr.1: 62819303-62819303 on Build GRCh38
rs17123872 Chr.1: 62819159-62819159 on Build GRCh38
rs571972655 Chr.1: 62819233-62819233 on Build GRCh38
rs746249675 Chr.1: 62819256-62819256 on Build GRCh38
rs746075279 Chr.1: 62819213-62819213 on Build GRCh38
rs895365337 Chr.1: 62819177-62819177 on Build GRCh38
rs763039923 Chr.1: 62819306-62819306 on Build GRCh38
rs3737891 Chr.1: 62819362-62819362 on Build GRCh38
rs566304475 Chr.1: 62819322-62819322 on Build GRCh38
rs201768764 Chr.1: 62819328-62819328 on Build GRCh38
rs765159860 Chr.1: 62819301-62819301 on Build GRCh38
rs777740309 Chr.1: 62819160-62819160 on Build GRCh38
rs747492130 Chr.1: 62819348-62819349 on Build GRCh38
rs762040130 Chr.1: 62819277-62819277 on Build GRCh38
rs542267992 Chr.1: 62819243-62819243 on Build GRCh38

View Full Product Details