Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153703960-153704191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAP31
Entrez Gene ID
10134
Gene Name
B-cell receptor-associated protein 31
Gene Aliases
6C6-AG, BAP31, CDM, DDCH, DXS1357E
UniGene
Hs.522817
Gene Chromosome Location
Chr.X: 153700492-153724746 on Build GRCh38
Transcripts
NM_001139441.1 NM_001139457.2 NM_001256447.1 NM_005745.7

SNP Detail

SNP ID Chromosome Location
rs782531748 Chr.X: 153704007-153704007 on Build GRCh38
rs368433664 Chr.X: 153704104-153704104 on Build GRCh38
rs373953555 Chr.X: 153704049-153704049 on Build GRCh38
rs782125326 Chr.X: 153704075-153704075 on Build GRCh38
rs782714259 Chr.X: 153704124-153704124 on Build GRCh38
rs781860748 Chr.X: 153704059-153704059 on Build GRCh38
rs11537731 Chr.X: 153704031-153704031 on Build GRCh38
rs371892645 Chr.X: 153704138-153704138 on Build GRCh38
rs150827588 Chr.X: 153704052-153704052 on Build GRCh38
rs781792281 Chr.X: 153704133-153704133 on Build GRCh38
rs781905415 Chr.X: 153703999-153703999 on Build GRCh38
rs111450526 Chr.X: 153704053-153704053 on Build GRCh38
rs782366321 Chr.X: 153704083-153704083 on Build GRCh38
rs781961537 Chr.X: 153704098-153704098 on Build GRCh38
rs199876065 Chr.X: 153704038-153704038 on Build GRCh38
rs11537730 Chr.X: 153704055-153704055 on Build GRCh38
rs782427826 Chr.X: 153704064-153704064 on Build GRCh38

View Full Product Details