Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 18514775-18515046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNB2
Entrez Gene ID
783
Gene Name
calcium voltage-gated channel auxiliary subunit beta 2
Gene Aliases
CACNLB2, CAVB2, MYSB
UniGene
Hs.59093
Gene Chromosome Location
Chr.10: 18140560-18545881 on Build GRCh38
Transcripts
NM_000724.3 NM_001167945.1 NM_201570.2 NM_201571.3 NM_201572.3 NM_201590.2 NM_201593.2 NM_201596.2 NM_201597.2 XM_005252588.3 XM_005252591.3 XM_006717502.3 XM_011519659.2 XM_011519660.2 XM_017016625.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7209808 c.695delA p.D232fs*31 Chr.10: 18515004-18515004 on Build GRCh38
210949 c.729T>G p.F243L Chr.10: 18515038-18515038 on Build GRCh38
259031 c.725G>A p.R242Q Chr.10: 18515034-18515034 on Build GRCh38
6486472 c.712G>A p.G238S Chr.10: 18515021-18515021 on Build GRCh38
3806884 c.710C>T p.A237V Chr.10: 18515019-18515019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142904006 Chr.10: 18514994-18514994 on Build GRCh38
rs532438177 Chr.10: 18514822-18514822 on Build GRCh38
rs777550976 Chr.10: 18514929-18514930 on Build GRCh38
rs772588058 Chr.10: 18514972-18514972 on Build GRCh38
rs947917992 Chr.10: 18514894-18514895 on Build GRCh38
rs1027424438 Chr.10: 18514982-18514982 on Build GRCh38
rs78902236 Chr.10: 18514862-18514862 on Build GRCh38
rs974378611 Chr.10: 18514965-18514965 on Build GRCh38
rs878969993 Chr.10: 18514827-18514827 on Build GRCh38
rs779482293 Chr.10: 18514939-18514939 on Build GRCh38
rs544599499 Chr.10: 18514858-18514858 on Build GRCh38
rs1019239354 Chr.10: 18514908-18514908 on Build GRCh38
rs570047550 Chr.10: 18514859-18514859 on Build GRCh38
rs577293026 Chr.10: 18514805-18514805 on Build GRCh38
rs549525106 Chr.10: 18514970-18514970 on Build GRCh38
rs920405057 Chr.10: 18514888-18514888 on Build GRCh38
rs771387497 Chr.10: 18515004-18515004 on Build GRCh38
rs1045333784 Chr.10: 18514905-18514905 on Build GRCh38
rs955165005 Chr.10: 18514820-18514820 on Build GRCh38
rs769349533 Chr.10: 18514932-18514932 on Build GRCh38
rs906445455 Chr.10: 18514922-18514923 on Build GRCh38
rs751262431 Chr.10: 18514957-18514960 on Build GRCh38
rs142899184 Chr.10: 18515022-18515022 on Build GRCh38
rs1037804431 Chr.10: 18514875-18514875 on Build GRCh38
rs777063041 Chr.10: 18515005-18515005 on Build GRCh38
rs1052092149 Chr.10: 18514980-18514980 on Build GRCh38

View Full Product Details