Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81550368-81550593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAAP100
Entrez Gene ID
80233
Gene Name
Fanconi anemia core complex associated protein 100
Gene Aliases
C17orf70
UniGene
Hs.313905
Gene Chromosome Location
Chr.17: 81539885-81552403 on Build GRCh38
Transcripts
NM_025161.5 NR_033338.1 XM_006722111.2 XM_006722112.3 XM_017025162.1 XM_017025163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4580249 c.618C>T p.R206R Chr.17: 81550423-81550423 on Build GRCh38
6082547 c.564G>A p.R188R Chr.17: 81550477-81550477 on Build GRCh38
6614725 c.1094A>G p.D365G Chr.17: 81550400-81550400 on Build GRCh38
2805262 c.1008C>T p.P336P Chr.17: 81550486-81550486 on Build GRCh38
6082544 c.923A>G p.H308R Chr.17: 81550571-81550571 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774395143 Chr.17: 81550535-81550535 on Build GRCh38
rs765745159 Chr.17: 81550428-81550428 on Build GRCh38
rs772355872 Chr.17: 81550423-81550423 on Build GRCh38
rs566124890 Chr.17: 81550478-81550478 on Build GRCh38
rs200471017 Chr.17: 81550434-81550434 on Build GRCh38
rs371626488 Chr.17: 81550402-81550402 on Build GRCh38
rs372168452 Chr.17: 81550425-81550425 on Build GRCh38
rs770311062 Chr.17: 81550531-81550532 on Build GRCh38
rs202147413 Chr.17: 81550429-81550429 on Build GRCh38
rs149199568 Chr.17: 81550456-81550456 on Build GRCh38
rs757431809 Chr.17: 81550547-81550547 on Build GRCh38
rs759987273 Chr.17: 81550426-81550426 on Build GRCh38
rs754908784 Chr.17: 81550446-81550446 on Build GRCh38
rs941450792 Chr.17: 81550515-81550515 on Build GRCh38
rs779001780 Chr.17: 81550447-81550447 on Build GRCh38
rs748803666 Chr.17: 81550422-81550423 on Build GRCh38
rs776922945 Chr.17: 81550480-81550480 on Build GRCh38
rs747224229 Chr.17: 81550477-81550477 on Build GRCh38
rs755921407 Chr.17: 81550394-81550394 on Build GRCh38
rs758308663 Chr.17: 81550539-81550539 on Build GRCh38
rs781386242 Chr.17: 81550567-81550567 on Build GRCh38
rs750106295 Chr.17: 81550386-81550386 on Build GRCh38
rs748165235 Chr.17: 81550450-81550450 on Build GRCh38
rs111682181 Chr.17: 81550540-81550540 on Build GRCh38
rs561678941 Chr.17: 81550407-81550407 on Build GRCh38
rs759401424 Chr.17: 81550524-81550524 on Build GRCh38
rs370963530 Chr.17: 81550420-81550420 on Build GRCh38
rs763497527 Chr.17: 81550496-81550496 on Build GRCh38
rs1022383889 Chr.17: 81550436-81550436 on Build GRCh38
rs916663738 Chr.17: 81550513-81550513 on Build GRCh38
rs753623012 Chr.17: 81550509-81550509 on Build GRCh38
rs746234663 Chr.17: 81550485-81550485 on Build GRCh38
rs891453308 Chr.17: 81550452-81550452 on Build GRCh38
rs766162023 Chr.17: 81550438-81550438 on Build GRCh38
rs533599529 Chr.17: 81550503-81550503 on Build GRCh38
rs772625798 Chr.17: 81550501-81550501 on Build GRCh38
rs368906561 Chr.17: 81550435-81550435 on Build GRCh38
rs771576578 Chr.17: 81550500-81550500 on Build GRCh38
rs915066414 Chr.17: 81550405-81550405 on Build GRCh38
rs368375305 Chr.17: 81550424-81550424 on Build GRCh38
rs753711909 Chr.17: 81550441-81550441 on Build GRCh38
rs751603864 Chr.17: 81550542-81550542 on Build GRCh38
rs761305579 Chr.17: 81550431-81550431 on Build GRCh38
rs779089599 Chr.17: 81550417-81550417 on Build GRCh38
rs755758038 Chr.17: 81550437-81550437 on Build GRCh38
rs201500435 Chr.17: 81550486-81550486 on Build GRCh38
rs373684364 Chr.17: 81550573-81550573 on Build GRCh38
rs779758461 Chr.17: 81550399-81550399 on Build GRCh38
rs777765031 Chr.17: 81550470-81550470 on Build GRCh38
rs139839925 Chr.17: 81550487-81550487 on Build GRCh38
rs748255197 Chr.17: 81550421-81550421 on Build GRCh38
rs765965102 Chr.17: 81550508-81550508 on Build GRCh38
rs755000149 Chr.17: 81550412-81550412 on Build GRCh38

View Full Product Details