Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144842873-144843110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD5
Entrez Gene ID
28991
Gene Name
COMM domain containing 5
Gene Aliases
HCARG, HT002
UniGene
Hs.493218
Gene Chromosome Location
Chr.8: 144837978-144853578 on Build GRCh38
Transcripts
NM_001282795.1 NM_001282796.1 NM_001282797.1 NM_003416.3 XM_006716654.2 XM_006716656.3 XM_011517291.2 XM_011517292.2 XM_011517293.2 XM_011517294.2 XM_011517295.2 XM_011517296.2 XM_011517297.2 XM_017013813.1 XM_017013814.1 XM_017013815.1 XM_017013816.1 XM_017013817.1

Gene Symbol
ZNF7
Entrez Gene ID
7553
Gene Name
zinc finger protein 7
Gene Aliases
HF.16, KOX4, zf30
UniGene
Hs.493218
Gene Chromosome Location
Chr.8: 144827500-144848070 on Build GRCh38
Transcripts
NM_001282795.1 NM_001282796.1 NM_001282797.1 NM_003416.3 XM_006716654.2 XM_006716656.3 XM_011517291.2 XM_011517292.2 XM_011517293.2 XM_011517294.2 XM_011517295.2 XM_011517296.2 XM_011517297.2 XM_017013813.1 XM_017013814.1 XM_017013815.1 XM_017013816.1 XM_017013817.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6047925 c.1791T>C p.I597I Chr.8: 144842898-144842898 on Build GRCh38
1097329 c.1780T>G p.S594A Chr.8: 144842887-144842887 on Build GRCh38
6832530 c.1971C>T p.T657T Chr.8: 144843078-144843078 on Build GRCh38
1569132 c.1871G>A p.R624H Chr.8: 144842978-144842978 on Build GRCh38
3646698 c.1852G>A p.G618R Chr.8: 144842959-144842959 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145249149 Chr.8: 144843078-144843078 on Build GRCh38
rs150360233 Chr.8: 144842975-144842975 on Build GRCh38
rs749763735 Chr.8: 144842994-144842994 on Build GRCh38
rs754477142 Chr.8: 144842997-144842997 on Build GRCh38
rs143638999 Chr.8: 144843075-144843075 on Build GRCh38
rs79367774 Chr.8: 144843007-144843007 on Build GRCh38
rs1020114581 Chr.8: 144843068-144843068 on Build GRCh38
rs780700459 Chr.8: 144842938-144842938 on Build GRCh38
rs899176056 Chr.8: 144842951-144842951 on Build GRCh38
rs767890130 Chr.8: 144843063-144843063 on Build GRCh38
rs781549082 Chr.8: 144842928-144842928 on Build GRCh38
rs370485110 Chr.8: 144843079-144843079 on Build GRCh38
rs1050831781 Chr.8: 144842909-144842909 on Build GRCh38
rs1735170 Chr.8: 144842893-144842893 on Build GRCh38
rs771768452 Chr.8: 144842956-144842956 on Build GRCh38
rs750306247 Chr.8: 144843043-144843043 on Build GRCh38
rs758471628 Chr.8: 144842899-144842899 on Build GRCh38
rs865993142 Chr.8: 144843080-144843080 on Build GRCh38
rs761791224 Chr.8: 144843059-144843059 on Build GRCh38
rs747797568 Chr.8: 144842946-144842946 on Build GRCh38
rs780753448 Chr.8: 144842992-144842992 on Build GRCh38
rs762586524 Chr.8: 144843022-144843023 on Build GRCh38
rs769903907 Chr.8: 144842965-144842965 on Build GRCh38
rs1039348382 Chr.8: 144842953-144842953 on Build GRCh38
rs747770085 Chr.8: 144843008-144843008 on Build GRCh38
rs750649043 Chr.8: 144843064-144843064 on Build GRCh38
rs765255445 Chr.8: 144842980-144842980 on Build GRCh38
rs776575162 Chr.8: 144843030-144843030 on Build GRCh38
rs748846039 Chr.8: 144842939-144842939 on Build GRCh38
rs778420738 Chr.8: 144843004-144843004 on Build GRCh38
rs777900708 Chr.8: 144842908-144842908 on Build GRCh38
rs992697304 Chr.8: 144843048-144843048 on Build GRCh38
rs760451269 Chr.8: 144842960-144842960 on Build GRCh38
rs757589476 Chr.8: 144842925-144842925 on Build GRCh38
rs562365113 Chr.8: 144843022-144843022 on Build GRCh38
rs9004 Chr.8: 144842937-144842937 on Build GRCh38
rs759448540 Chr.8: 144843033-144843033 on Build GRCh38
rs773897225 Chr.8: 144842945-144842945 on Build GRCh38
rs771816122 Chr.8: 144843015-144843015 on Build GRCh38
rs756569220 Chr.8: 144843074-144843074 on Build GRCh38
rs372906310 Chr.8: 144842897-144842897 on Build GRCh38
rs772935138 Chr.8: 144842958-144842958 on Build GRCh38
rs149536664 Chr.8: 144843044-144843044 on Build GRCh38
rs1004969475 Chr.8: 144842904-144842904 on Build GRCh38
rs751787394 Chr.8: 144842923-144842923 on Build GRCh38
rs762995729 Chr.8: 144842974-144842974 on Build GRCh38
rs766373722 Chr.8: 144842963-144842963 on Build GRCh38
rs138064022 Chr.8: 144842977-144842977 on Build GRCh38
rs757438179 Chr.8: 144842978-144842978 on Build GRCh38
rs756514326 Chr.8: 144842985-144842985 on Build GRCh38
rs746849807 Chr.8: 144843025-144843025 on Build GRCh38
rs768190813 Chr.8: 144842942-144842942 on Build GRCh38

View Full Product Details