Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 98208243-98208516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
R3HCC1L
Entrez Gene ID
27291
Gene Name
R3H domain and coiled-coil containing 1 like
Gene Aliases
C10orf28, GIDRP86, GIDRP88, PSORT
UniGene
Hs.419800
Gene Chromosome Location
Chr.10: 98134608-98244897 on Build GRCh38
Transcripts
NM_001256619.1 NM_001256620.1 NM_001256621.1 NM_014472.4 NM_138469.2 XM_011539639.1 XM_011539640.1 XM_011539641.1 XM_011539642.1 XM_011539643.1 XM_011539644.1 XM_011539645.2 XM_011539646.1 XM_011539647.1 XM_011539648.1 XM_011539649.1 XM_017016074.1 XM_017016075.1 XM_017016076.1 XM_017016077.1 XM_017016078.1 XM_017016079.1 XM_017016080.1 XM_017016081.1 XM_017016082.1 XM_017016083.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3442463 c.205C>T p.R69* Chr.10: 98208319-98208319 on Build GRCh38
4666505 c.173delA p.E60fs*12 Chr.10: 98208287-98208287 on Build GRCh38
1188017 c.334G>T p.E112* Chr.10: 98208448-98208448 on Build GRCh38
3442461 c.167T>A p.L56H Chr.10: 98208281-98208281 on Build GRCh38
4484833 c.285C>A p.D95E Chr.10: 98208399-98208399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761439395 Chr.10: 98208282-98208282 on Build GRCh38
rs775931614 Chr.10: 98208443-98208443 on Build GRCh38
rs768383441 Chr.10: 98208280-98208280 on Build GRCh38
rs143631828 Chr.10: 98208338-98208338 on Build GRCh38
rs765832966 Chr.10: 98208300-98208300 on Build GRCh38
rs1019720514 Chr.10: 98208377-98208377 on Build GRCh38
rs761155634 Chr.10: 98208390-98208390 on Build GRCh38
rs758583644 Chr.10: 98208324-98208324 on Build GRCh38
rs775591805 Chr.10: 98208278-98208278 on Build GRCh38
rs555004057 Chr.10: 98208296-98208296 on Build GRCh38
rs769525398 Chr.10: 98208362-98208362 on Build GRCh38
rs754242171 Chr.10: 98208398-98208398 on Build GRCh38
rs373657280 Chr.10: 98208320-98208320 on Build GRCh38
rs764870906 Chr.10: 98208284-98208284 on Build GRCh38
rs780159397 Chr.10: 98208327-98208327 on Build GRCh38
rs758679632 Chr.10: 98208433-98208433 on Build GRCh38
rs762292516 Chr.10: 98208403-98208403 on Build GRCh38
rs750722213 Chr.10: 98208428-98208428 on Build GRCh38
rs372164414 Chr.10: 98208385-98208385 on Build GRCh38
rs1003266861 Chr.10: 98208274-98208274 on Build GRCh38
rs754229437 Chr.10: 98208313-98208313 on Build GRCh38
rs746975266 Chr.10: 98208328-98208328 on Build GRCh38
rs754950643 Chr.10: 98208482-98208482 on Build GRCh38
rs772878995 Chr.10: 98208368-98208368 on Build GRCh38
rs753188303 Chr.10: 98208303-98208303 on Build GRCh38
rs776472833 Chr.10: 98208281-98208281 on Build GRCh38
rs868106160 Chr.10: 98208452-98208452 on Build GRCh38
rs757629684 Chr.10: 98208318-98208318 on Build GRCh38
rs755899792 Chr.10: 98208276-98208276 on Build GRCh38
rs751664482 Chr.10: 98208454-98208454 on Build GRCh38
rs781208585 Chr.10: 98208487-98208487 on Build GRCh38
rs762559219 Chr.10: 98208376-98208376 on Build GRCh38
rs753163228 Chr.10: 98208420-98208421 on Build GRCh38
rs747984030 Chr.10: 98208353-98208353 on Build GRCh38
rs772789148 Chr.10: 98208293-98208293 on Build GRCh38
rs761224574 Chr.10: 98208311-98208311 on Build GRCh38
rs7903980 Chr.10: 98208408-98208408 on Build GRCh38
rs370226522 Chr.10: 98208337-98208337 on Build GRCh38
rs996651942 Chr.10: 98208466-98208466 on Build GRCh38
rs779291841 Chr.10: 98208319-98208319 on Build GRCh38
rs766101897 Chr.10: 98208343-98208346 on Build GRCh38
rs374768883 Chr.10: 98208425-98208425 on Build GRCh38
rs764428816 Chr.10: 98208312-98208312 on Build GRCh38
rs770480067 Chr.10: 98208380-98208380 on Build GRCh38
rs12775148 Chr.10: 98208451-98208451 on Build GRCh38
rs7918729 Chr.10: 98208409-98208409 on Build GRCh38
rs780064819 Chr.10: 98208440-98208440 on Build GRCh38
rs764667669 Chr.10: 98208393-98208393 on Build GRCh38

View Full Product Details