Product Details
- Primer Sequences
- Log in to view
- Primer Chromosome Location
-
Chr.1:
145868147-145867896
on Build GRCh38
View in UCSC browser
- Format
- Each primer will be delivered in a single tube, dried-down
- Quick Reference Guide
- Primer Designer™ Tool Quick Reference
- Ampliseq ID's
Gene Details
- Gene Symbol
- ANKRD35
- Entrez Gene ID
-
148741
- Gene Name
- ankyrin repeat domain 35
- Gene Aliases
-
-
- UniGene
- Hs.710624
- Gene Chromosome Location
-
Chr.1:
145866560-145885866
on Build GRCh38
- Transcripts
-
NM_001280799.1
NM_144698.4
Cosmic Detail
Cosmic ID |
Mutation |
AA |
Transcript Location |
4135558 |
c.2924A>G |
p.H975R |
Chr.1:
145868010-145868010
on Build GRCh38
|
3984171 |
c.2923C>T |
p.H975Y |
Chr.1:
145868011-145868011
on Build GRCh38
|
1736717 |
c.2878G>A |
p.D960N |
Chr.1:
145868056-145868056
on Build GRCh38
|
View Full Product Details